Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007713.hg.1 |
ENST00000558899.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007713.hg.1 |
ENST00000558899.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007713.hg.1 |
ENST00000558899.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007713.hg.1 |
ENST00000558899.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007713.hg.1 |
ENST00000558899.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007713.hg.1 |
ENST00000614698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007713.hg.1 |
ENST00000614698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007713.hg.1 |
NR_110007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007713.hg.1 |
NR_110007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007713.hg.1 |
NR_110007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007713.hg.1 |
NR_110007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007713.hg.1 |
NR_110007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007713.hg.1 |
NR_110007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007713.hg.1 |
NR_110008.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007713.hg.1 |
NR_110008.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007713.hg.1 |
NR_110008.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007713.hg.1 |
NR_110008.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007713.hg.1 |
NR_110008.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007713.hg.1 |
NR_110009.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007713.hg.1 |
NR_110009.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007713.hg.1 |
NR_110009.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007713.hg.1 |
NR_110009.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007713.hg.1 |
NR_110009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007713.hg.1 |
ENST00000558899.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007713.hg.1 |
ENST00000558899.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007713.hg.1 |
ENST00000558899.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007713.hg.1 |
ENST00000558899.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007713.hg.1 |
ENST00000558899.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007713.hg.1 |
ENST00000614698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007713.hg.1 |
ENST00000614698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007713.hg.1 |
NR_110007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007713.hg.1 |
NR_110007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007713.hg.1 |
NR_110007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007713.hg.1 |
NR_110007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007713.hg.1 |
NR_110007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007713.hg.1 |
NR_110007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007713.hg.1 |
NR_110008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007713.hg.1 |
NR_110008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007713.hg.1 |
NR_110008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007713.hg.1 |
NR_110008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007713.hg.1 |
NR_110008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007713.hg.1 |
NR_110009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007713.hg.1 |
NR_110009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007713.hg.1 |
NR_110009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007713.hg.1 |
NR_110009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007713.hg.1 |
NR_110009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000153746.hg.1 |
ENST00000558899.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000153746.hg.1 |
ENST00000614698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000153746.hg.1 |
NR_110007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000153746.hg.1 |
NR_110008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000153746.hg.1 |
NR_110009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000153746.hg.1 |
TC2000007713.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000153746.hg.1 |
ENST00000558899.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000153746.hg.1 |
ENST00000614698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000153746.hg.1 |
NR_110007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000153746.hg.1 |
NR_110008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000153746.hg.1 |
NR_110009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000153737.hg.1 |
ENST00000558899.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000153737.hg.1 |
NR_110007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000153737.hg.1 |
NR_110008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000153737.hg.1 |
NR_110009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000153737.hg.1 |
TC2000007713.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000153737.hg.1 |
ENST00000558899.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000153737.hg.1 |
NR_110007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000153737.hg.1 |
NR_110008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000153737.hg.1 |
NR_110009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000153742.hg.1 |
ENST00000558899.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000153742.hg.1 |
ENST00000614698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000153742.hg.1 |
NR_110007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000153742.hg.1 |
NR_110008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000153742.hg.1 |
NR_110009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000153742.hg.1 |
TC2000007713.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000153742.hg.1 |
ENST00000558899.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000153742.hg.1 |
ENST00000614698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000153742.hg.1 |
NR_110007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000153742.hg.1 |
NR_110008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000153742.hg.1 |
NR_110009.1 |
Refseq |