Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011544.hg.1 |
ENST00000550287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011544.hg.1 |
ENST00000550287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011544.hg.1 |
ENST00000550287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011544.hg.1 |
ENST00000551107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011544.hg.1 |
ENST00000551107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011544.hg.1 |
NR_110092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011544.hg.1 |
NR_110092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011544.hg.1 |
NR_110092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011544.hg.1 |
NR_110093.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011544.hg.1 |
NR_110093.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011544.hg.1 |
NR_110093.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011544.hg.1 |
ENST00000550287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011544.hg.1 |
ENST00000550287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011544.hg.1 |
ENST00000550287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011544.hg.1 |
ENST00000551107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011544.hg.1 |
ENST00000551107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011544.hg.1 |
NR_110092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011544.hg.1 |
NR_110092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011544.hg.1 |
NR_110092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011544.hg.1 |
NR_110093.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011544.hg.1 |
NR_110093.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011544.hg.1 |
NR_110093.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200189467.hg.1 |
ENST00000551107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189467.hg.1 |
NR_110093.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200189467.hg.1 |
TC1200011544.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200189467.hg.1 |
NR_110093.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200189467.hg.1 |
ENST00000551107.1 |
Ensembl |