Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008440.hg.1 |
ENST00000421483.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008440.hg.1 |
ENST00000421483.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008440.hg.1 |
ENST00000421483.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008440.hg.1 |
ENST00000421483.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008440.hg.1 |
ENST00000421483.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008440.hg.1 |
ENST00000421483.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008440.hg.1 |
ENST00000441399.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008440.hg.1 |
ENST00000441399.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008440.hg.1 |
ENST00000441399.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008440.hg.1 |
ENST00000441399.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008440.hg.1 |
ENST00000441399.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008440.hg.1 |
ENST00000441399.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008440.hg.1 |
NR_110410.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008440.hg.1 |
NR_110410.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008440.hg.1 |
NR_110410.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008440.hg.1 |
NR_110410.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008440.hg.1 |
NR_110410.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00157445.hg.1 |
ENST00000441399.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00157445.hg.1 |
NR_110410.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00157445.hg.1 |
TC0X00008440.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00157445.hg.1 |
ENST00000441399.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00157445.hg.1 |
NR_110410.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00157449.hg.1 |
ENST00000421483.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00157449.hg.1 |
ENST00000421483.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00157449.hg.1 |
ENST00000441399.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00157449.hg.1 |
NR_110410.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00157449.hg.1 |
TC0X00008440.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00157449.hg.1 |
ENST00000421483.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00157449.hg.1 |
ENST00000421483.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00157449.hg.1 |
ENST00000441399.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00157449.hg.1 |
NR_110410.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008440.hg.1 |
ENST00000421483.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008440.hg.1 |
ENST00000421483.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008440.hg.1 |
ENST00000421483.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008440.hg.1 |
ENST00000421483.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008440.hg.1 |
ENST00000421483.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008440.hg.1 |
ENST00000421483.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008440.hg.1 |
ENST00000441399.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008440.hg.1 |
ENST00000441399.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008440.hg.1 |
ENST00000441399.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008440.hg.1 |
ENST00000441399.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008440.hg.1 |
ENST00000441399.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008440.hg.1 |
ENST00000441399.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008440.hg.1 |
NR_110410.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008440.hg.1 |
NR_110410.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008440.hg.1 |
NR_110410.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008440.hg.1 |
NR_110410.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008440.hg.1 |
NR_110410.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00157444.hg.1 |
ENST00000441399.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00157444.hg.1 |
NR_110410.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00157444.hg.1 |
TC0X00008440.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00157444.hg.1 |
ENST00000441399.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00157444.hg.1 |
NR_110410.1 |
Refseq |