Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04404475_m1
          See other OTUD6B-AS1 GE Assays ›
          Gene Symbol:
          OTUD6B-AS1
          Gene Name:
          OTUD6B antisense RNA 1 (head to head)
          Gene Aliases:
          GS1-251I9.4
          Chromosome Location:
          Chr.8: 91059906 - 91070189 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04404475_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: OTUD6B-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_110438.1 2 - 3 500 139
          GenBank mRNA BM715864.1 - 2 - 3 434 139
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BI831785.1 - 122 Chr. - - [-] on Build 139
          GenBankmRNA AA919125.1 - 1 - 2 Chr. 8 - 91063512 - 91069987 [+] on Build GRCh38 139
          GenBankmRNA AA876258.1 - 1 - 2 Chr. 8 - 91063519 - 91069668 [+] on Build GRCh38 139
          GenBankmRNA AA993252.1 - 1 - 2 Chr. 8 - 91063519 - 91069732 [+] on Build GRCh38 139
          GenBankmRNA CB052236.1 - 1 - 2 Chr. 8 - 91063533 - 91069703 [+] on Build GRCh38 139
          GenBankmRNA AA625959.1 - 1 - 2 Chr. 8 - 91063517 - 91069723 [+] on Build GRCh38 139
          GenBankmRNA BG528387.1 - 2 - 3 433 Chr. 8 - 91063481 - 91070115 [-] on Build GRCh38 139
          GenBankmRNA BU569666.1 - 2 - 3 354 Chr. 8 - 91063510 - 91070043 [-] on Build GRCh38 139
          GenBankmRNA AA813069.1 - 1 - 2 Chr. 8 - 91063512 - 91069723 [+] on Build GRCh38 139
          GenBankmRNA DA121039.1 - 2 - 3 366 Chr. 8 - 91063571 - 91070054 [-] on Build GRCh38 139
          GenBankmRNA BG699467.1 - 398 Chr. - - [-] on Build 139
          GenBankmRNA DA317375.1 - 2 - 3 423 Chr. 8 - 91063596 - 91070112 [-] on Build GRCh38 139
          GenBankmRNA BU955253.1 - 367 Chr. - - [-] on Build 139
          GenBankmRNA AI052136.1 - 1 - 2 Chr. 8 - 91063512 - 91069946 [+] on Build GRCh38 139
          GenBankmRNA DA812941.1 - 2 - 3 350 Chr. 8 - 91063280 - 91070038 [-] on Build GRCh38 139
          GenBankmRNA AA861352.1 - 1 - 2 Chr. 8 - 91063519 - 91069694 [+] on Build GRCh38 139
          GenBankmRNA BQ011370.1 - 1 - 2 Chr. 8 - 91063512 - 91069801 [+] on Build GRCh38 139
          GenBankmRNA AI123542.1 - 1 - 2 Chr. 8 - 91063517 - 91069788 [+] on Build GRCh38 139
          GenBankmRNA DA094126.1 - 2 - 3 334 Chr. 8 - 91063521 - 91070023 [-] on Build GRCh38 139
          GenBankmRNA HY047845.1 - 2 - 3 350 Chr. 8 - 91063649 - 91070039 [-] on Build GRCh38 139
          GenBankmRNA BM674193.1 - 1 - 2 Chr. 8 - 91063512 - 91070105 [+] on Build GRCh38 139
          GenBankmRNA AV725083.1 - 2 - 3 Chr. 8 - 91063522 - 91070021 [-] on Build GRCh38 139
          GenBankmRNA BM792763.1 - 1 - 2 151 Chr. 8 - 91063519 - 91069732 [-] on Build GRCh38 139
          GenBankmRNA AI289651.1 - 1 - 2 Chr. 8 - 91063519 - 91069754 [+] on Build GRCh38 139
          GenBankmRNA AI458129.1 - 1 - 2 Chr. 8 - 91063512 - 91069974 [+] on Build GRCh38 139
          GenBankmRNA BI824111.1 - 2 - 3 373 Chr. 8 - 91063397 - 91070059 [-] on Build GRCh38 139

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream transmembrane protein 55A TMEM55A -18.9kb Sense Chr. 8 - 90994271 - 91040975 on NCBI Build GRCh38
          Downstream OTU domain containing 6B OTUD6B CGI-77,
          DUBA-5,
          DUBA5
          374.0 Anti-sense Chr. 8 - 91070196 - 91087095 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0800011077.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0800011077.hg.1 ENST00000522817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800011077.hg.1 ENST00000522817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800011077.hg.1 ENST00000522817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800011077.hg.1 ENST00000522817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800011077.hg.1 ENST00000522817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800011077.hg.1 ENST00000522817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800011077.hg.1 ENST00000522817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800011077.hg.1 ENST00000522817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800011077.hg.1 ENST00000524003.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800011077.hg.1 ENST00000524003.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800011077.hg.1 ENST00000524003.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800011077.hg.1 ENST00000524003.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800011077.hg.1 ENST00000524003.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800011077.hg.1 ENST00000524003.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800011077.hg.1 NR_110438.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800011077.hg.1 NR_110438.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800011077.hg.1 NR_110438.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800011077.hg.1 NR_110438.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800011077.hg.1 NR_110438.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800011077.hg.1 NR_110438.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800011077.hg.1 NR_110438.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800011077.hg.1 NR_110438.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800011077.hg.1 NR_110438.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800011077.hg.1 NR_110439.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800011077.hg.1 NR_110439.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800011077.hg.1 NR_110439.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800011077.hg.1 NR_110439.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800011077.hg.1 NR_110439.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800011077.hg.1 NR_110439.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800175360.hg.1 ENST00000522817.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800175360.hg.1 NR_110438.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800175360.hg.1 TC0800011077.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800175360.hg.1 NR_110438.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800175360.hg.1 ENST00000522817.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800175363.hg.1 ENST00000522817.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800175363.hg.1 NR_110438.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800175363.hg.1 TC0800011077.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800175363.hg.1 ENST00000522817.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800175363.hg.1 NR_110438.1 Refseq
          Affymetrix Human Clariom D Assay TC0800011077.hg.1 ENST00000522817.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800011077.hg.1 ENST00000522817.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800011077.hg.1 ENST00000522817.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800011077.hg.1 ENST00000522817.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800011077.hg.1 ENST00000522817.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800011077.hg.1 ENST00000522817.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800011077.hg.1 ENST00000522817.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800011077.hg.1 ENST00000522817.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800011077.hg.1 ENST00000524003.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800011077.hg.1 ENST00000524003.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800011077.hg.1 ENST00000524003.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800011077.hg.1 ENST00000524003.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800011077.hg.1 ENST00000524003.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800011077.hg.1 ENST00000524003.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800011077.hg.1 NR_110438.1 Refseq
          Affymetrix Human Clariom D Assay TC0800011077.hg.1 NR_110438.1 Refseq
          Affymetrix Human Clariom D Assay TC0800011077.hg.1 NR_110438.1 Refseq
          Affymetrix Human Clariom D Assay TC0800011077.hg.1 NR_110438.1 Refseq
          Affymetrix Human Clariom D Assay TC0800011077.hg.1 NR_110438.1 Refseq
          Affymetrix Human Clariom D Assay TC0800011077.hg.1 NR_110438.1 Refseq
          Affymetrix Human Clariom D Assay TC0800011077.hg.1 NR_110438.1 Refseq
          Affymetrix Human Clariom D Assay TC0800011077.hg.1 NR_110438.1 Refseq
          Affymetrix Human Clariom D Assay TC0800011077.hg.1 NR_110438.1 Refseq
          Affymetrix Human Clariom D Assay TC0800011077.hg.1 NR_110439.1 Refseq
          Affymetrix Human Clariom D Assay TC0800011077.hg.1 NR_110439.1 Refseq
          Affymetrix Human Clariom D Assay TC0800011077.hg.1 NR_110439.1 Refseq
          Affymetrix Human Clariom D Assay TC0800011077.hg.1 NR_110439.1 Refseq
          Affymetrix Human Clariom D Assay TC0800011077.hg.1 NR_110439.1 Refseq
          Affymetrix Human Clariom D Assay TC0800011077.hg.1 NR_110439.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800175361.hg.1 ENST00000522817.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800175361.hg.1 NR_110438.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800175361.hg.1 TC0800011077.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800175361.hg.1 ENST00000522817.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800175361.hg.1 NR_110438.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800175362.hg.1 ENST00000522817.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800175362.hg.1 NR_110438.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800175362.hg.1 TC0800011077.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800175362.hg.1 ENST00000522817.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800175362.hg.1 NR_110438.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800175364.hg.1 ENST00000522817.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800175364.hg.1 NR_110438.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800175364.hg.1 TC0800011077.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800175364.hg.1 ENST00000522817.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800175364.hg.1 NR_110438.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0