Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016620.hg.1 |
ENST00000420221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016620.hg.1 |
ENST00000420221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016620.hg.1 |
ENST00000420221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016620.hg.1 |
NR_110580.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016620.hg.1 |
NR_110580.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016620.hg.1 |
NR_110580.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016620.hg.1 |
NR_110580.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016620.hg.1 |
NR_110580.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016620.hg.1 |
NR_110580.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200225543.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200225543.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200225543.hg.1 |
NR_110580.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200225543.hg.1 |
TC0200016620.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200225543.hg.1 |
NR_110580.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200225543.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200225543.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200225545.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200225545.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200225545.hg.1 |
NR_110580.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200225545.hg.1 |
TC0200016620.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200225545.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200225545.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200225545.hg.1 |
NR_110580.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016620.hg.1 |
ENST00000420221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016620.hg.1 |
ENST00000420221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016620.hg.1 |
ENST00000420221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016620.hg.1 |
ENST00000453678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016620.hg.1 |
NR_110580.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016620.hg.1 |
NR_110580.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016620.hg.1 |
NR_110580.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016620.hg.1 |
NR_110580.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016620.hg.1 |
NR_110580.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016620.hg.1 |
NR_110580.1 |
Refseq |