Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007842.hg.1 |
ENST00000626308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007842.hg.1 |
ENST00000626308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007842.hg.1 |
ENST00000626308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007842.hg.1 |
ENST00000627052.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007842.hg.1 |
ENST00000627052.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007842.hg.1 |
ENST00000627052.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007842.hg.1 |
ENST00000627052.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010058.hg.1 |
ENST00000625254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010058.hg.1 |
ENST00000625254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010058.hg.1 |
ENST00000625254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010058.hg.1 |
ENST00000625254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010058.hg.1 |
ENST00000625287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010058.hg.1 |
ENST00000625287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010058.hg.1 |
ENST00000625287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010058.hg.1 |
ENST00000625287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010058.hg.1 |
ENST00000625287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010058.hg.1 |
ENST00000625369.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010058.hg.1 |
ENST00000625369.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010058.hg.1 |
ENST00000625369.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010058.hg.1 |
ENST00000625369.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010058.hg.1 |
ENST00000626729.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010058.hg.1 |
ENST00000626729.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010058.hg.1 |
ENST00000626729.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010058.hg.1 |
ENST00000626729.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010058.hg.1 |
ENST00000626729.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010058.hg.1 |
ENST00000626729.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010058.hg.1 |
ENST00000626729.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010058.hg.1 |
ENST00000630421.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010058.hg.1 |
ENST00000630421.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010058.hg.1 |
ENST00000630421.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010058.hg.1 |
ENST00000630421.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010058.hg.1 |
NR_110731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010058.hg.1 |
NR_110731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010058.hg.1 |
NR_110731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010058.hg.1 |
NR_110731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010058.hg.1 |
NR_110731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010058.hg.1 |
NR_110731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010058.hg.1 |
NR_110731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300168018.hg.1 |
ENST00000625254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300168018.hg.1 |
ENST00000625369.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300168018.hg.1 |
NR_110731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300168018.hg.1 |
TC1300010058.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300168018.hg.1 |
NR_110731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300168018.hg.1 |
ENST00000625254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300168018.hg.1 |
ENST00000625369.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300168019.hg.1 |
ENST00000625254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300168019.hg.1 |
ENST00000625369.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300168019.hg.1 |
NR_110731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300168019.hg.1 |
TC1300010058.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300168019.hg.1 |
ENST00000625254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300168019.hg.1 |
ENST00000625369.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300168019.hg.1 |
NR_110731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010058.hg.1 |
ENST00000625254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010058.hg.1 |
ENST00000625254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010058.hg.1 |
ENST00000625254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010058.hg.1 |
ENST00000625254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010058.hg.1 |
ENST00000625287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010058.hg.1 |
ENST00000625287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010058.hg.1 |
ENST00000625287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010058.hg.1 |
ENST00000625287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010058.hg.1 |
ENST00000625287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010058.hg.1 |
ENST00000625369.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010058.hg.1 |
ENST00000625369.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010058.hg.1 |
ENST00000625369.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010058.hg.1 |
ENST00000625369.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010058.hg.1 |
ENST00000626729.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010058.hg.1 |
ENST00000626729.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010058.hg.1 |
ENST00000626729.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010058.hg.1 |
ENST00000626729.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010058.hg.1 |
ENST00000626729.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010058.hg.1 |
ENST00000626729.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010058.hg.1 |
ENST00000626729.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010058.hg.1 |
ENST00000630421.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010058.hg.1 |
ENST00000630421.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010058.hg.1 |
ENST00000630421.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010058.hg.1 |
ENST00000630421.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010058.hg.1 |
NR_110731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010058.hg.1 |
NR_110731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010058.hg.1 |
NR_110731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010058.hg.1 |
NR_110731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010058.hg.1 |
NR_110731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010058.hg.1 |
NR_110731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010058.hg.1 |
NR_110731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300168020.hg.1 |
ENST00000625254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300168020.hg.1 |
NR_110731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300168020.hg.1 |
TC1300010058.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300168020.hg.1 |
ENST00000625254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300168020.hg.1 |
NR_110731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007842.hg.1 |
ENST00000626308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007842.hg.1 |
ENST00000626308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007842.hg.1 |
ENST00000626308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007842.hg.1 |
ENST00000627052.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007842.hg.1 |
ENST00000627052.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007842.hg.1 |
ENST00000627052.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007842.hg.1 |
ENST00000627052.2 |
Ensembl |