Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04405370_m1
          See other SVIL-AS1 GE Assays ›
          Gene Symbol:
          SVIL-AS1
          Gene Name:
          SVIL antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 29409534 - 29487856 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04405370_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SVIL-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003930.2 1 - 2 123 78
          NR_110922.1 1 - 2 123 78
          NR_110923.1 1 - 2 123 78
          NR_110926.1 1 - 2 123 78
          NR_110927.1 1 - 2 123 78
          NR_110929.1 1 - 2 123 78
          GenBank mRNA BC031656.1 - 1 - 2 78 78
          BC066966.1 - 1 - 2 121 78
          BF739922.1 - 2 - 3 78
          BX461223.2 - 1 - 2 31 78
          DA024320.1 - 1 - 2 123 78
          DA745252.1 - 1 - 2 68 78
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AV710390.1 - 1 - 2 99 Chr. 10 - 29409558 - 29483504 [+] on Build GRCh38 78
          GenBankmRNA DA802966.1 - 1 - 2 100 Chr. 10 - 29409557 - 29510818 [+] on Build GRCh38 78
          GenBankmRNA DA121252.1 - 1 - 2 99 Chr. 10 - 29409558 - 29483221 [+] on Build GRCh38 78
          GenBankmRNA DA661027.1 - 1 - 2 121 Chr. 10 - 29409536 - 29420878 [+] on Build GRCh38 78
          GenBankmRNA DA581920.1 - 1 - 2 100 Chr. 10 - 29409557 - 29420885 [+] on Build GRCh38 78
          GenBankmRNA HY001946.1 - 1 - 2 101 Chr. 10 - 29409556 - 29454522 [+] on Build GRCh38 78
          GenBankmRNA DA177235.1 - 1 - 2 68 Chr. 10 - 29409589 - 29420684 [+] on Build GRCh38 78
          GenBankmRNA DB180401.1 - 1 - 2 119 Chr. 10 - 29409538 - 29420869 [+] on Build GRCh38 78
          GenBankmRNA W56452.1 - 1 - 2 256 Chr. 10 - 29409402 - 29458400 [+] on Build GRCh38 78
          GenBankmRNA DB262217.1 - 1 - 2 100 Chr. 10 - 29409557 - 29420897 [+] on Build GRCh38 78
          GenBankmRNA DA322935.1 - 1 - 2 123 Chr. 10 - 29409534 - 29420775 [+] on Build GRCh38 78
          GenBankmRNA DB047449.1 - 1 - 2 93 Chr. 10 - 29409564 - 29420900 [+] on Build GRCh38 78
          GenBankmRNA CB960614.1 - 1 - 2 127 Chr. 10 - 29409557 - 29415958 [+] on Build GRCh38 78
          GenBankmRNA DA507912.1 - 1 - 2 100 Chr. 10 - 29409557 - 29483252 [+] on Build GRCh38 78
          GenBankmRNA DA888030.1 - 1 - 2 100 Chr. 10 - 29409557 - 29420895 [+] on Build GRCh38 78
          GenBankmRNA DA265970.1 - 1 - 2 94 Chr. 10 - 29409564 - 29487550 [+] on Build GRCh38 78
          GenBankmRNA DB099389.1 - 1 - 2 123 Chr. 10 - 29409534 - 29420897 [+] on Build GRCh38 78
          GenBankmRNA DB049688.1 - 1 - 2 93 Chr. 10 - 29409564 - 29420901 [+] on Build GRCh38 78
          GenBankmRNA DB031339.1 - 1 - 2 121 Chr. 10 - 29409537 - 29420901 [+] on Build GRCh38 78
          GenBankmRNA DA307490.1 - 1 - 2 123 Chr. 10 - 29409534 - 29483247 [+] on Build GRCh38 78
          GenBankmRNA DA110605.1 - 1 - 2 123 Chr. 10 - 29409534 - 29470500 [+] on Build GRCh38 78
          GenBankmRNA BG036994.1 - 1 - 2 126 Chr. 10 - 29409539 - 29487715 [+] on Build GRCh38 78
          GenBankmRNA DA591198.1 - 1 - 2 123 Chr. 10 - 29409534 - 29420881 [+] on Build GRCh38 78
          GenBankmRNA DA373323.1 - 1 - 2 100 Chr. 10 - 29409557 - 29420883 [+] on Build GRCh38 78
          GenBankmRNA DA580038.1 - 1 - 2 93 Chr. 10 - 29409564 - 29420906 [+] on Build GRCh38 78
          GenBankmRNA DB284993.1 - 1 - 2 100 Chr. 10 - 29409557 - 29420877 [+] on Build GRCh38 78
          GenBankmRNA DA119810.1 - 1 - 2 110 Chr. 10 - 29409547 - 29458376 [+] on Build GRCh38 78
          GenBankmRNA BM565031.1 - 1 - 2 67 Chr. 10 - 29409597 - 29420946 [+] on Build GRCh38 78
          GenBankmRNA BI756656.1 - 1 - 2 85 Chr. 10 - 29409572 - 29420950 [+] on Build GRCh38 78
          GenBankmRNA DB012592.1 - 1 - 2 96 Chr. 10 - 29409561 - 29420899 [+] on Build GRCh38 78
          GenBankmRNA DA168383.1 - 1 - 2 123 Chr. 10 - 29409534 - 29420880 [+] on Build GRCh38 78
          GenBankmRNA CB988224.1 - 1 - 2 127 Chr. 10 - 29409557 - 29421050 [+] on Build GRCh38 78
          GenBankmRNA BM511177.1 - 1 - 2 67 Chr. 10 - 29409597 - 29420778 [+] on Build GRCh38 78
          GenBankmRNA DA449336.1 - 1 - 2 123 Chr. 10 - 29409534 - 29420863 [+] on Build GRCh38 78
          GenBankmRNA CB046493.1 - 1 - 2 59 Chr. 10 - 29409607 - 29458607 [+] on Build GRCh38 78
          GenBankmRNA DB003348.1 - 1 - 2 123 Chr. 10 - 29409534 - 29420847 [+] on Build GRCh38 78
          GenBankmRNA DA836156.1 - 1 - 2 58 Chr. 10 - 29409599 - 29420931 [+] on Build GRCh38 78
          GenBankmRNA DB167786.1 - 1 - 2 68 Chr. 10 - 29409589 - 29483291 [+] on Build GRCh38 78
          GenBankmRNA BM835655.1 - 1 - 2 80 Chr. 10 - 29409577 - 29420879 [+] on Build GRCh38 78
          GenBankmRNA DB056630.1 - 1 - 2 123 Chr. 10 - 29409534 - 29420764 [+] on Build GRCh38 78
          GenBankmRNA DA177718.1 - 1 - 2 120 Chr. 10 - 29409538 - 29487538 [+] on Build GRCh38 78
          GenBankmRNA CK823803.1 - 1 - 2 67 Chr. 10 - 29409597 - 29420937 [+] on Build GRCh38 78
          GenBankmRNA DA570840.1 - 581 Chr. - - [-] on Build 78
          GenBankmRNA H38002.1 - 1 - 2 33 Chr. 10 - 29409624 - 29483353 [+] on Build GRCh38 78
          GenBankmRNA DA536274.1 - 1 - 2 68 Chr. 10 - 29409589 - 29415881 [+] on Build GRCh38 78
          GenBankmRNA DA548063.1 - 1 - 2 123 Chr. 10 - 29409534 - 29420868 [+] on Build GRCh38 78
          GenBankmRNA DB042440.1 - 1 - 2 93 Chr. 10 - 29409564 - 29420876 [+] on Build GRCh38 78
          GenBankmRNA HY158995.1 - 1 - 2 98 Chr. 10 - 29409559 - 29458600 [+] on Build GRCh38 78
          GenBankmRNA DA405633.1 - 1 - 2 119 Chr. 10 - 29409538 - 29420863 [+] on Build GRCh38 78
          GenBankmRNA DA831992.1 - 1 - 2 94 Chr. 10 - 29409564 - 29421166 [+] on Build GRCh38 78
          GenBankmRNA DB091461.1 - 1 - 2 119 Chr. 10 - 29409538 - 29420869 [+] on Build GRCh38 78
          GenBankmRNA BI820719.1 - 1 - 2 77 Chr. 10 - 29409580 - 29421127 [+] on Build GRCh38 78
          GenBankmRNA DA003665.1 - 1 - 2 93 Chr. 10 - 29409564 - 29420919 [+] on Build GRCh38 78
          GenBankmRNA BI916268.1 - 1 - 2 66 Chr. 10 - 29409591 - 29483451 [+] on Build GRCh38 78

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream lysozyme like 1 LYZL1 KAAG648,
          LYC2,
          LYZD1,
          PRO1278,
          bA534G20.1
          -91.2kb Sense Chr. 10 - 29289051 - 29318328 on NCBI Build GRCh38
          Downstream supervillin SVIL 41.9kb Anti-sense Chr. 10 - 29457338 - 29737001 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0