Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008406.hg.1 |
ENST00000555255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008406.hg.1 |
ENST00000555255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008406.hg.1 |
ENST00000555255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008406.hg.1 |
ENST00000555364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008406.hg.1 |
ENST00000555364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008406.hg.1 |
ENST00000555364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008406.hg.1 |
ENST00000555364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008406.hg.1 |
ENST00000555364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008406.hg.1 |
ENST00000555364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008406.hg.1 |
ENST00000555364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008406.hg.1 |
ENST00000555364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008406.hg.1 |
ENST00000555364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008406.hg.1 |
ENST00000555364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008406.hg.1 |
NR_120322.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008406.hg.1 |
NR_120322.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008406.hg.1 |
NR_120322.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008406.hg.1 |
NR_120322.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008406.hg.1 |
NR_120322.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008406.hg.1 |
NR_120322.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008406.hg.1 |
NR_120323.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008406.hg.1 |
NR_120323.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008406.hg.1 |
NR_120323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500161010.hg.1 |
ENST00000555255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500161010.hg.1 |
NR_120323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500161010.hg.1 |
TC1500008406.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500161010.hg.1 |
NR_120323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500161010.hg.1 |
ENST00000555255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008406.hg.1 |
ENST00000555255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008406.hg.1 |
ENST00000555255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008406.hg.1 |
ENST00000555255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008406.hg.1 |
ENST00000555364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008406.hg.1 |
ENST00000555364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008406.hg.1 |
ENST00000555364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008406.hg.1 |
ENST00000555364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008406.hg.1 |
ENST00000555364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008406.hg.1 |
ENST00000555364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008406.hg.1 |
ENST00000555364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008406.hg.1 |
ENST00000555364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008406.hg.1 |
ENST00000555364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008406.hg.1 |
ENST00000555364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008406.hg.1 |
NR_120322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008406.hg.1 |
NR_120322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008406.hg.1 |
NR_120322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008406.hg.1 |
NR_120322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008406.hg.1 |
NR_120322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008406.hg.1 |
NR_120322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008406.hg.1 |
NR_120323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008406.hg.1 |
NR_120323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008406.hg.1 |
NR_120323.1 |
Refseq |