Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008784.hg.1 |
ENST00000439938.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008784.hg.1 |
ENST00000439938.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008784.hg.1 |
ENST00000439938.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008784.hg.1 |
ENST00000439938.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008784.hg.1 |
ENST00000439938.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008784.hg.1 |
ENST00000439938.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008784.hg.1 |
NR_120343.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008784.hg.1 |
NR_120343.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008784.hg.1 |
NR_120343.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008784.hg.1 |
NR_120343.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008784.hg.1 |
NR_120343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008784.hg.1 |
ENST00000439938.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008784.hg.1 |
ENST00000439938.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008784.hg.1 |
ENST00000439938.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008784.hg.1 |
ENST00000439938.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008784.hg.1 |
ENST00000439938.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008784.hg.1 |
ENST00000439938.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008784.hg.1 |
NR_120343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008784.hg.1 |
NR_120343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008784.hg.1 |
NR_120343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008784.hg.1 |
NR_120343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008784.hg.1 |
NR_120343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500163452.hg.1 |
ENST00000439938.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500163452.hg.1 |
NR_120343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500163452.hg.1 |
TC1500008784.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500163452.hg.1 |
NR_120343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500163452.hg.1 |
ENST00000439938.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500163459.hg.1 |
ENST00000439938.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500163459.hg.1 |
NR_120343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500163459.hg.1 |
TC1500008784.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500163459.hg.1 |
ENST00000439938.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500163459.hg.1 |
NR_120343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500163457.hg.1 |
ENST00000439938.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500163457.hg.1 |
NR_120343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500163457.hg.1 |
TC1500008784.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500163457.hg.1 |
ENST00000439938.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500163457.hg.1 |
NR_120343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500163458.hg.1 |
ENST00000439938.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500163458.hg.1 |
NR_120343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500163458.hg.1 |
TC1500008784.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500163458.hg.1 |
ENST00000439938.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500163458.hg.1 |
NR_120343.1 |
Refseq |