Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010362.hg.1 |
ENST00000506090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010362.hg.1 |
ENST00000506090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010362.hg.1 |
ENST00000506090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010362.hg.1 |
ENST00000506090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010362.hg.1 |
ENST00000506090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010362.hg.1 |
ENST00000506090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010362.hg.1 |
ENST00000506090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010362.hg.1 |
ENST00000506090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010362.hg.1 |
ENST00000506090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010362.hg.1 |
ENST00000506090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010362.hg.1 |
ENST00000558835.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010362.hg.1 |
ENST00000558835.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010362.hg.1 |
ENST00000560368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010362.hg.1 |
ENST00000560368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010362.hg.1 |
ENST00000560368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010362.hg.1 |
ENST00000560368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010362.hg.1 |
NR_120369.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010362.hg.1 |
NR_120369.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010362.hg.1 |
NR_120369.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010362.hg.1 |
NR_120369.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010362.hg.1 |
NR_120369.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010362.hg.1 |
NR_120370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010362.hg.1 |
NR_120370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010362.hg.1 |
NR_120370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010362.hg.1 |
NR_120370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010362.hg.1 |
NR_120370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500178017.hg.1 |
ENST00000506090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500178017.hg.1 |
ENST00000506090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500178017.hg.1 |
ENST00000560368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500178017.hg.1 |
NR_120369.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500178017.hg.1 |
NR_120370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500178017.hg.1 |
TC1500010362.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500178017.hg.1 |
NR_120369.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500178017.hg.1 |
ENST00000506090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500178017.hg.1 |
ENST00000506090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500178017.hg.1 |
ENST00000560368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500178017.hg.1 |
NR_120370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500178029.hg.1 |
ENST00000560368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500178029.hg.1 |
NR_120370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500178029.hg.1 |
TC1500010362.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500178029.hg.1 |
ENST00000560368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500178029.hg.1 |
NR_120370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500178028.hg.1 |
ENST00000560368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500178028.hg.1 |
NR_120370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500178028.hg.1 |
TC1500010362.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500178028.hg.1 |
ENST00000560368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500178028.hg.1 |
NR_120370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010362.hg.1 |
ENST00000506090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010362.hg.1 |
ENST00000506090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010362.hg.1 |
ENST00000506090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010362.hg.1 |
ENST00000506090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010362.hg.1 |
ENST00000506090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010362.hg.1 |
ENST00000506090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010362.hg.1 |
ENST00000506090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010362.hg.1 |
ENST00000506090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010362.hg.1 |
ENST00000506090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010362.hg.1 |
ENST00000506090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010362.hg.1 |
ENST00000558835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010362.hg.1 |
ENST00000558835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010362.hg.1 |
ENST00000560368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010362.hg.1 |
ENST00000560368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010362.hg.1 |
ENST00000560368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010362.hg.1 |
ENST00000560368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010362.hg.1 |
NR_120369.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010362.hg.1 |
NR_120369.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010362.hg.1 |
NR_120369.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010362.hg.1 |
NR_120369.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010362.hg.1 |
NR_120369.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010362.hg.1 |
NR_120370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010362.hg.1 |
NR_120370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010362.hg.1 |
NR_120370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010362.hg.1 |
NR_120370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010362.hg.1 |
NR_120370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500178016.hg.1 |
ENST00000506090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500178016.hg.1 |
ENST00000506090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500178016.hg.1 |
NR_120369.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500178016.hg.1 |
NR_120370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500178016.hg.1 |
TC1500010362.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500178016.hg.1 |
ENST00000506090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500178016.hg.1 |
ENST00000506090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500178016.hg.1 |
NR_120369.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500178016.hg.1 |
NR_120370.1 |
Refseq |