Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013533.hg.1 |
ENST00000517726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013533.hg.1 |
ENST00000517726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013533.hg.1 |
ENST00000517726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013533.hg.1 |
ENST00000517726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013533.hg.1 |
ENST00000519050.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013533.hg.1 |
ENST00000519050.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013533.hg.1 |
ENST00000519050.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013533.hg.1 |
ENST00000519050.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013533.hg.1 |
ENST00000519218.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013533.hg.1 |
ENST00000519218.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013533.hg.1 |
ENST00000519218.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013533.hg.1 |
NR_120507.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013533.hg.1 |
NR_120507.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013533.hg.1 |
NR_120507.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013533.hg.1 |
NR_120507.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700200233.hg.1 |
ENST00000517726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700200233.hg.1 |
ENST00000519050.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700200233.hg.1 |
ENST00000519218.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700200233.hg.1 |
NR_120507.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700200233.hg.1 |
TC0700013533.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700200233.hg.1 |
ENST00000517726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700200233.hg.1 |
ENST00000519050.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700200233.hg.1 |
ENST00000519218.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700200233.hg.1 |
NR_120507.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700200238.hg.1 |
ENST00000517726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700200238.hg.1 |
NR_120507.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700200238.hg.1 |
TC0700013533.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700200238.hg.1 |
ENST00000517726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700200238.hg.1 |
NR_120507.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700200239.hg.1 |
ENST00000517726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700200239.hg.1 |
NR_120507.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700200239.hg.1 |
TC0700013533.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700200239.hg.1 |
ENST00000517726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700200239.hg.1 |
NR_120507.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013533.hg.1 |
ENST00000517726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013533.hg.1 |
ENST00000517726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013533.hg.1 |
ENST00000517726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013533.hg.1 |
ENST00000517726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013533.hg.1 |
ENST00000519050.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013533.hg.1 |
ENST00000519050.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013533.hg.1 |
ENST00000519050.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013533.hg.1 |
ENST00000519050.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013533.hg.1 |
ENST00000519218.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013533.hg.1 |
ENST00000519218.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013533.hg.1 |
ENST00000519218.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013533.hg.1 |
NR_120507.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013533.hg.1 |
NR_120507.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013533.hg.1 |
NR_120507.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013533.hg.1 |
NR_120507.1 |
Refseq |