Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007580.hg.1 |
ENST00000426205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007580.hg.1 |
ENST00000426205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007580.hg.1 |
ENST00000426205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007580.hg.1 |
ENST00000426205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007580.hg.1 |
ENST00000458615.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007580.hg.1 |
ENST00000458615.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007580.hg.1 |
ENST00000458615.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007580.hg.1 |
ENST00000458615.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007580.hg.1 |
ENST00000458615.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007580.hg.1 |
ENST00000458615.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007580.hg.1 |
ENST00000458615.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007580.hg.1 |
ENST00000458615.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007580.hg.1 |
ENST00000458615.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007580.hg.1 |
ENST00000458615.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007580.hg.1 |
ENST00000458615.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007580.hg.1 |
ENST00000458615.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007580.hg.1 |
NR_120515.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007580.hg.1 |
NR_120515.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007580.hg.1 |
NR_120515.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007580.hg.1 |
NR_120515.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007580.hg.1 |
NR_120515.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007580.hg.1 |
NR_120515.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007580.hg.1 |
NR_120515.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007580.hg.1 |
NR_120516.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007580.hg.1 |
NR_120516.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007580.hg.1 |
NR_120516.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007580.hg.1 |
NR_120516.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007580.hg.1 |
NR_120516.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007580.hg.1 |
NR_120516.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007580.hg.1 |
ENST00000426205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007580.hg.1 |
ENST00000426205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007580.hg.1 |
ENST00000426205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007580.hg.1 |
ENST00000426205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007580.hg.1 |
ENST00000458615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007580.hg.1 |
ENST00000458615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007580.hg.1 |
ENST00000458615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007580.hg.1 |
ENST00000458615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007580.hg.1 |
ENST00000458615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007580.hg.1 |
ENST00000458615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007580.hg.1 |
ENST00000458615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007580.hg.1 |
ENST00000458615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007580.hg.1 |
ENST00000458615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007580.hg.1 |
ENST00000458615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007580.hg.1 |
ENST00000458615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007580.hg.1 |
ENST00000458615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007580.hg.1 |
NR_120515.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007580.hg.1 |
NR_120515.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007580.hg.1 |
NR_120515.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007580.hg.1 |
NR_120515.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007580.hg.1 |
NR_120515.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007580.hg.1 |
NR_120515.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007580.hg.1 |
NR_120515.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007580.hg.1 |
NR_120516.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007580.hg.1 |
NR_120516.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007580.hg.1 |
NR_120516.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007580.hg.1 |
NR_120516.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007580.hg.1 |
NR_120516.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007580.hg.1 |
NR_120516.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700152519.hg.1 |
ENST00000426205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700152519.hg.1 |
NR_120515.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700152519.hg.1 |
TC0700007580.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700152519.hg.1 |
NR_120515.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700152519.hg.1 |
ENST00000426205.1 |
Ensembl |