Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007314.hg.1 |
ENST00000510551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007314.hg.1 |
ENST00000510551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007314.hg.1 |
ENST00000510551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007314.hg.1 |
ENST00000510551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007314.hg.1 |
ENST00000510551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007314.hg.1 |
ENST00000510551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007314.hg.1 |
ENST00000510551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007314.hg.1 |
NR_121640.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007314.hg.1 |
NR_121640.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007314.hg.1 |
NR_121640.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007314.hg.1 |
NR_121640.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007314.hg.1 |
NR_121640.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007314.hg.1 |
NR_121640.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007314.hg.1 |
NR_121640.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007314.hg.1 |
NR_121641.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007314.hg.1 |
NR_121641.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007314.hg.1 |
NR_121641.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007314.hg.1 |
NR_121641.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007314.hg.1 |
NR_121641.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150048.hg.1 |
NR_121641.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150048.hg.1 |
TC0400007314.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400150048.hg.1 |
NR_121641.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150057.hg.1 |
ENST00000510551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150057.hg.1 |
NR_121640.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150057.hg.1 |
NR_121641.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150057.hg.1 |
TC0400007314.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400150057.hg.1 |
ENST00000510551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150057.hg.1 |
NR_121640.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150057.hg.1 |
NR_121641.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150046.hg.1 |
NR_121641.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150046.hg.1 |
TC0400007314.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400150046.hg.1 |
NR_121641.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150056.hg.1 |
ENST00000510551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150056.hg.1 |
NR_121640.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150056.hg.1 |
NR_121641.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150056.hg.1 |
TC0400007314.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400150056.hg.1 |
ENST00000510551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150056.hg.1 |
NR_121640.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150056.hg.1 |
NR_121641.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007314.hg.1 |
ENST00000510551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007314.hg.1 |
ENST00000510551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007314.hg.1 |
ENST00000510551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007314.hg.1 |
ENST00000510551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007314.hg.1 |
ENST00000510551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007314.hg.1 |
ENST00000510551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007314.hg.1 |
ENST00000510551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007314.hg.1 |
NR_121640.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007314.hg.1 |
NR_121640.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007314.hg.1 |
NR_121640.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007314.hg.1 |
NR_121640.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007314.hg.1 |
NR_121640.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007314.hg.1 |
NR_121640.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007314.hg.1 |
NR_121640.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007314.hg.1 |
NR_121641.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007314.hg.1 |
NR_121641.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007314.hg.1 |
NR_121641.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007314.hg.1 |
NR_121641.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007314.hg.1 |
NR_121641.1 |
Refseq |