Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008390.hg.1 |
ENST00000465249.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008390.hg.1 |
ENST00000465249.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008390.hg.1 |
ENST00000465249.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008390.hg.1 |
ENST00000465249.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008390.hg.1 |
ENST00000627586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008390.hg.1 |
ENST00000627586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008390.hg.1 |
ENST00000627586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008390.hg.1 |
ENST00000627586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008390.hg.1 |
ENST00000627586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008390.hg.1 |
ENST00000628886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008390.hg.1 |
ENST00000628886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008390.hg.1 |
ENST00000628886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008390.hg.1 |
ENST00000628886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008390.hg.1 |
NR_121661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008390.hg.1 |
NR_121661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008390.hg.1 |
NR_121661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008390.hg.1 |
NR_121661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008390.hg.1 |
ENST00000465249.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008390.hg.1 |
ENST00000465249.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008390.hg.1 |
ENST00000465249.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008390.hg.1 |
ENST00000465249.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008390.hg.1 |
ENST00000627586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008390.hg.1 |
ENST00000627586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008390.hg.1 |
ENST00000627586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008390.hg.1 |
ENST00000627586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008390.hg.1 |
ENST00000627586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008390.hg.1 |
ENST00000628886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008390.hg.1 |
ENST00000628886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008390.hg.1 |
ENST00000628886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008390.hg.1 |
ENST00000628886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008390.hg.1 |
NR_121661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008390.hg.1 |
NR_121661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008390.hg.1 |
NR_121661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008390.hg.1 |
NR_121661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300159992.hg.1 |
ENST00000465249.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300159992.hg.1 |
ENST00000628886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300159992.hg.1 |
NR_121661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300159992.hg.1 |
TC0300008390.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300159992.hg.1 |
NR_121661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300159992.hg.1 |
ENST00000465249.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300159992.hg.1 |
ENST00000628886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300159991.hg.1 |
ENST00000465249.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300159991.hg.1 |
ENST00000628886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300159991.hg.1 |
NR_121661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300159991.hg.1 |
TC0300008390.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300159991.hg.1 |
ENST00000465249.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300159991.hg.1 |
ENST00000628886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300159991.hg.1 |
NR_121661.1 |
Refseq |