Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013132.hg.1 |
ENST00000416982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013132.hg.1 |
ENST00000416982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013132.hg.1 |
ENST00000416982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013132.hg.1 |
ENST00000416982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013132.hg.1 |
ENST00000416982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013132.hg.1 |
ENST00000416982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013132.hg.1 |
ENST00000453187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013132.hg.1 |
ENST00000453187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013132.hg.1 |
ENST00000454441.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013132.hg.1 |
ENST00000454441.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013132.hg.1 |
ENST00000454441.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013132.hg.1 |
ENST00000454441.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013132.hg.1 |
ENST00000454441.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013132.hg.1 |
NR_125776.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013132.hg.1 |
NR_125776.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013132.hg.1 |
NR_125776.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013132.hg.1 |
NR_125776.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013132.hg.1 |
NR_125776.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194770.hg.1 |
ENST00000454441.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194770.hg.1 |
NR_125776.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194770.hg.1 |
TC0700013132.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700194770.hg.1 |
NR_125776.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194770.hg.1 |
ENST00000454441.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013132.hg.1 |
ENST00000416982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013132.hg.1 |
ENST00000416982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013132.hg.1 |
ENST00000416982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013132.hg.1 |
ENST00000416982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013132.hg.1 |
ENST00000416982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013132.hg.1 |
ENST00000416982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013132.hg.1 |
ENST00000453187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013132.hg.1 |
ENST00000453187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013132.hg.1 |
ENST00000454441.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013132.hg.1 |
ENST00000454441.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013132.hg.1 |
ENST00000454441.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013132.hg.1 |
ENST00000454441.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013132.hg.1 |
ENST00000454441.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013132.hg.1 |
NR_125776.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013132.hg.1 |
NR_125776.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013132.hg.1 |
NR_125776.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013132.hg.1 |
NR_125776.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013132.hg.1 |
NR_125776.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194775.hg.1 |
ENST00000416982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194775.hg.1 |
ENST00000454441.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194775.hg.1 |
NR_125776.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194775.hg.1 |
TC0700013132.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700194775.hg.1 |
ENST00000416982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194775.hg.1 |
ENST00000454441.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194775.hg.1 |
NR_125776.1 |
Refseq |