Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000512674.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000512674.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000512674.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000512674.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000512674.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000512674.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000512674.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000512674.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000512674.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000608785.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000608785.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000608785.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000608785.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000608785.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000610223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000610223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000610223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000610223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000610223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000610223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000610223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000610683.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000610683.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000610683.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000610683.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000610683.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000610683.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000610683.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000617323.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000617323.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000617323.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000617323.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000617323.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000617323.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000617323.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000617323.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
ENST00000617323.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
NR_125933.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
NR_125933.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
NR_125933.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
NR_125933.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
NR_125933.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
NR_125933.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
NR_125933.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
NR_125933.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
NR_125933.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
NR_125934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
NR_125934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
NR_125934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
NR_125934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
NR_125934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
NR_125934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
NR_125934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012604.hg.1 |
NR_125934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400186213.hg.1 |
ENST00000512674.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186213.hg.1 |
ENST00000610223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186213.hg.1 |
ENST00000610683.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186213.hg.1 |
ENST00000617323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186213.hg.1 |
NR_125933.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400186213.hg.1 |
NR_125934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400186213.hg.1 |
TC0400012604.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400186213.hg.1 |
ENST00000610683.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186213.hg.1 |
ENST00000512674.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186213.hg.1 |
ENST00000610223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186213.hg.1 |
ENST00000617323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186213.hg.1 |
NR_125933.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400186213.hg.1 |
NR_125934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000512674.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000512674.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000512674.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000512674.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000512674.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000512674.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000512674.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000512674.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000512674.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000608785.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000608785.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000608785.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000608785.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000608785.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000610223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000610223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000610223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000610223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000610223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000610223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000610223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000610683.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000610683.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000610683.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000610683.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000610683.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000610683.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000610683.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000617323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000617323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000617323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000617323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000617323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000617323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000617323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000617323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
ENST00000617323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
NR_125933.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
NR_125933.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
NR_125933.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
NR_125933.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
NR_125933.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
NR_125933.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
NR_125933.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
NR_125933.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
NR_125933.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
NR_125934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
NR_125934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
NR_125934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
NR_125934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
NR_125934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
NR_125934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
NR_125934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012604.hg.1 |
NR_125934.1 |
Refseq |