Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008497.hg.1 |
ENST00000592917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008497.hg.1 |
ENST00000592917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008497.hg.1 |
ENST00000592917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008497.hg.1 |
ENST00000592917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008497.hg.1 |
NR_126009.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008497.hg.1 |
NR_126009.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008497.hg.1 |
NR_126009.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008497.hg.1 |
NR_126009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700163320.hg.1 |
ENST00000592917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700163320.hg.1 |
NR_126009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700163320.hg.1 |
TC1700008497.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700163320.hg.1 |
NR_126009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700163320.hg.1 |
ENST00000592917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008497.hg.1 |
ENST00000592917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008497.hg.1 |
ENST00000592917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008497.hg.1 |
ENST00000592917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008497.hg.1 |
ENST00000592917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008497.hg.1 |
NR_126009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008497.hg.1 |
NR_126009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008497.hg.1 |
NR_126009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008497.hg.1 |
NR_126009.1 |
Refseq |