Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012197.hg.1 |
ENST00000456514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012197.hg.1 |
ENST00000456514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012197.hg.1 |
ENST00000456514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012197.hg.1 |
ENST00000456514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012197.hg.1 |
NR_073453.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012197.hg.1 |
NR_073453.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012197.hg.1 |
NR_073453.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012197.hg.1 |
NR_073453.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000183376.hg.1 |
ENST00000456514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183376.hg.1 |
NR_073453.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000183376.hg.1 |
TC1000012197.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000183376.hg.1 |
NR_073453.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000183376.hg.1 |
ENST00000456514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012197.hg.1 |
ENST00000456514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012197.hg.1 |
ENST00000456514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012197.hg.1 |
ENST00000456514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012197.hg.1 |
ENST00000456514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012197.hg.1 |
NR_073453.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012197.hg.1 |
NR_073453.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012197.hg.1 |
NR_073453.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012197.hg.1 |
NR_073453.1 |
Refseq |