Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_103806.1 | 1 - 2 | 116 | 104 | ||
GenBank mRNA | BC043243.2 | - | 1 - 2 | 85 | 104 | |
HY004923.1 | - | 1 - 2 | 117 | 104 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | inner mitochondrial membrane peptidase subunit 2 | IMMP2L |
IMMP2L-IT1, IMP2, IMP2-LIKE |
-246.0kb | Anti-sense | Chr. 7 - 110662645 - 111562531 on NCBI Build GRCh38 |
Downstream | zinc finger protein 277 | ZNF277 |
NRIF4, ZNF277P |
384.8kb | Sense | Chr. 7 - 112206588 - 112343934 on NCBI Build GRCh38 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |