Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0200183100.hg.1 |
ENST00000356150.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200183100.hg.1 |
ENST00000405430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200183100.hg.1 |
ENST00000463865.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200183100.hg.1 |
ENST00000468321.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200183100.hg.1 |
ENST00000472012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200183100.hg.1 |
ENST00000626873.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200183100.hg.1 |
NM_001282687.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200183100.hg.1 |
NM_015677.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200183100.hg.1 |
NR_104223.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200183100.hg.1 |
NR_104225.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200183100.hg.1 |
TC0200011449.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200183100.hg.1 |
ENST00000356150.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200183100.hg.1 |
ENST00000405430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200183100.hg.1 |
ENST00000463865.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200183100.hg.1 |
ENST00000468321.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200183100.hg.1 |
ENST00000472012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200183100.hg.1 |
ENST00000626873.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200183100.hg.1 |
NM_001282687.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200183100.hg.1 |
NM_015677.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200183100.hg.1 |
NR_104223.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200183100.hg.1 |
NR_104225.1 |
Refseq |