Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006579.hg.1 |
ENST00000455199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006579.hg.1 |
ENST00000455199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006579.hg.1 |
ENST00000455199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006579.hg.1 |
ENST00000455199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006579.hg.1 |
ENST00000455199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006579.hg.1 |
ENST00000455199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006579.hg.1 |
ENST00000455199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006579.hg.1 |
ENST00000455199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006579.hg.1 |
ENST00000455525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006579.hg.1 |
ENST00000455525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006579.hg.1 |
NR_120632.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006579.hg.1 |
NR_120632.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006579.hg.1 |
NR_120632.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006579.hg.1 |
NR_120632.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006579.hg.1 |
NR_120632.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006579.hg.1 |
NR_120632.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006579.hg.1 |
NR_120632.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006579.hg.1 |
NR_120632.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000144995.hg.1 |
ENST00000455199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000144995.hg.1 |
NR_120632.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000144995.hg.1 |
TC1000006579.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000144995.hg.1 |
NR_120632.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000144995.hg.1 |
ENST00000455199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000144994.hg.1 |
ENST00000455199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000144994.hg.1 |
NR_120632.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000144994.hg.1 |
TC1000006579.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000144994.hg.1 |
ENST00000455199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000144994.hg.1 |
NR_120632.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006579.hg.1 |
ENST00000455199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006579.hg.1 |
ENST00000455199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006579.hg.1 |
ENST00000455199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006579.hg.1 |
ENST00000455199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006579.hg.1 |
ENST00000455199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006579.hg.1 |
ENST00000455199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006579.hg.1 |
ENST00000455199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006579.hg.1 |
ENST00000455199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006579.hg.1 |
ENST00000455525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006579.hg.1 |
ENST00000455525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006579.hg.1 |
NR_120632.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006579.hg.1 |
NR_120632.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006579.hg.1 |
NR_120632.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006579.hg.1 |
NR_120632.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006579.hg.1 |
NR_120632.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006579.hg.1 |
NR_120632.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006579.hg.1 |
NR_120632.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006579.hg.1 |
NR_120632.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000144996.hg.1 |
ENST00000455199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000144996.hg.1 |
NR_120632.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000144996.hg.1 |
TC1000006579.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000144996.hg.1 |
ENST00000455199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000144996.hg.1 |
NR_120632.1 |
Refseq |