Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006821.hg.1 |
ENST00000432626.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006821.hg.1 |
ENST00000432626.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006821.hg.1 |
NR_046638.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006821.hg.1 |
NR_046638.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146700.hg.1 |
ENST00000432626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146700.hg.1 |
NR_046638.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146700.hg.1 |
TC0X00006821.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00146700.hg.1 |
ENST00000432626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146700.hg.1 |
NR_046638.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146701.hg.1 |
ENST00000432626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146701.hg.1 |
NR_046638.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146701.hg.1 |
TC0X00006821.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00146701.hg.1 |
ENST00000432626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146701.hg.1 |
NR_046638.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006821.hg.1 |
ENST00000432626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006821.hg.1 |
ENST00000432626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006821.hg.1 |
NR_046638.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006821.hg.1 |
NR_046638.1 |
Refseq |