Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009827.hg.1 |
ENST00000360538.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009827.hg.1 |
ENST00000360538.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009827.hg.1 |
ENST00000360538.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009827.hg.1 |
ENST00000360538.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009827.hg.1 |
ENST00000360538.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009827.hg.1 |
ENST00000379858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009827.hg.1 |
ENST00000379858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009827.hg.1 |
ENST00000379858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009827.hg.1 |
NM_001195622.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009827.hg.1 |
NM_001195622.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009827.hg.1 |
NM_001195622.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009827.hg.1 |
NM_001195622.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009827.hg.1 |
NM_001195622.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009827.hg.1 |
NM_005802.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009827.hg.1 |
NM_005802.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009827.hg.1 |
NM_005802.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009827.hg.1 |
NM_005802.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009827.hg.1 |
NM_005802.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009827.hg.1 |
NM_005802.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009827.hg.1 |
ENST00000360538.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009827.hg.1 |
ENST00000360538.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009827.hg.1 |
ENST00000360538.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009827.hg.1 |
ENST00000360538.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009827.hg.1 |
ENST00000360538.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009827.hg.1 |
ENST00000379858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009827.hg.1 |
ENST00000379858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009827.hg.1 |
ENST00000379858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009827.hg.1 |
NM_001195622.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009827.hg.1 |
NM_001195622.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009827.hg.1 |
NM_001195622.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009827.hg.1 |
NM_001195622.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009827.hg.1 |
NM_001195622.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009827.hg.1 |
NM_005802.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009827.hg.1 |
NM_005802.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009827.hg.1 |
NM_005802.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009827.hg.1 |
NM_005802.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009827.hg.1 |
NM_005802.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009827.hg.1 |
NM_005802.4 |
Refseq |