Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs04935921_m1
See other TRIP12 GE Assays ›
Gene Symbol:
TRIP12
Gene Name:
thyroid hormone receptor interactor 12
Gene Aliases:
TRIP-12, ULF
Chromosome Location:
Chr.2: 229763837 - 229923234 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs04935921_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: TRIP12
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001284214.1
NP_001271143.1
2 - 3
278
79
XM_005246954.4
XP_005247011.1
2 - 3
302
79
XM_005246955.4
XP_005247012.1
2 - 3
301
79
XM_005246956.4
XP_005247013.1
2 - 3
281
79
XM_005246957.4
XP_005247014.1
2 - 3
268
79
XM_005246958.4
XP_005247015.1
2 - 3
302
79
XM_005246960.4
XP_005247017.1
2 - 3
302
79
XM_011512180.2
XP_011510482.1
2 - 3
255
79
XM_011512181.2
XP_011510483.1
2 - 3
253
79
XM_017005272.1
XP_016860761.1
3 - 4
361
79
XM_017005273.1
XP_016860762.1
2 - 3
228
79
XM_017005274.1
XP_016860763.1
2 - 3
176
79
XM_017005275.1
XP_016860764.1
2 - 3
234
79
XM_017005276.1
XP_016860765.1
2 - 3
239
79
XM_017005277.1
XP_016860766.1
3 - 4
379
79
XM_017005278.1
XP_016860767.1
2 - 3
194
79
XM_017005279.1
XP_016860768.1
2 - 3
266
79
XM_017005280.1
XP_016860769.1
2 - 3
255
79
XM_017005281.1
XP_016860770.1
2 - 3
255
79
XM_017005282.1
XP_016860771.1
2 - 3
301
79
XM_017005283.1
XP_016860772.1
2 - 3
302
79
XM_017005284.1
XP_016860773.1
2 - 3
302
79
XM_017005289.1
XP_016860778.1
2 - 3
302
79
XM_017005290.1
XP_016860779.1
2 - 3
302
79
GenBank mRNA
AK302468.1
-
2 - 3
232
79
BC114556.1
-
2 - 3
172
40083165
79
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
Mammalian Gene Collection
Homo sapiens
Probe spans exons
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
DNA repair
ubiquitin-dependent protein catabolic process
cellular response to DNA damage stimulus
embryo development
negative regulation of histone H2A K63-linked ubiquitination
negative regulation of double-strand break repair
ubiquitin-protein transferase activity
protein binding
ligase activity
thyroid hormone receptor binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0