Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013531.hg.1 |
ENST00000430375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013531.hg.1 |
ENST00000430375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013531.hg.1 |
ENST00000430375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013531.hg.1 |
ENST00000430375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013531.hg.1 |
ENST00000430375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013531.hg.1 |
ENST00000430375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016133.hg.1 |
ENST00000444926.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016133.hg.1 |
ENST00000444926.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016133.hg.1 |
ENST00000444926.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013531.hg.1 |
ENST00000430375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013531.hg.1 |
ENST00000430375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013531.hg.1 |
ENST00000430375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013531.hg.1 |
ENST00000430375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013531.hg.1 |
ENST00000430375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013531.hg.1 |
ENST00000430375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016133.hg.1 |
ENST00000444926.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016133.hg.1 |
ENST00000444926.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016133.hg.1 |
ENST00000444926.1 |
Ensembl |