Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010059.hg.1 |
ENST00000366259.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010059.hg.1 |
ENST00000366259.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007842.hg.1 |
ENST00000626308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007842.hg.1 |
ENST00000626308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007842.hg.1 |
ENST00000626308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007842.hg.1 |
ENST00000627052.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007842.hg.1 |
ENST00000627052.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007842.hg.1 |
ENST00000627052.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007842.hg.1 |
ENST00000627052.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007842.hg.1 |
ENST00000626308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007842.hg.1 |
ENST00000626308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007842.hg.1 |
ENST00000626308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007842.hg.1 |
ENST00000627052.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007842.hg.1 |
ENST00000627052.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007842.hg.1 |
ENST00000627052.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007842.hg.1 |
ENST00000627052.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010059.hg.1 |
ENST00000366259.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010059.hg.1 |
ENST00000366259.2 |
Ensembl |