Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04938231_m1
          See other LOC100506098 GE Assays ›
          Gene Symbol:
          LOC100506098
          Gene Name:
          uncharacterized LOC100506098
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 20217577 - 20221700 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04938231_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100506098
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_134567.1 4 - 5 489 62
          NR_134568.1 4 - 5 398 62
          NR_134569.1 3 - 4 327 62
          GenBank mRNA AI928513.1 - 1 - 2 62
          BG777800.1 - 3 - 4 328 62
          BI759100.1 - 4 - 5 389 62
          DR423379.1 - 4 - 5 474 62
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AA708705.1 - 1 - 2 Chr. 7 - 20219618 - 20221695 [-] on Build GRCh38 62
          GenBankmRNA BF114992.1 - 1 - 2 Chr. 7 - 20218343 - 20221697 [-] on Build GRCh38 62
          GenBankmRNA BE220080.1 - 1 - 2 Chr. 7 - 20219609 - 20221696 [-] on Build GRCh38 62
          GenBankmRNA AW242719.1 - 1 - 2 Chr. 7 - 20219611 - 20221697 [-] on Build GRCh38 62
          GenBankmRNA AI629030.1 - 1 - 2 Chr. 7 - 20219613 - 20221699 [-] on Build GRCh38 62
          GenBankmRNA AW151071.1 - 1 - 2 Chr. 7 - 20219613 - 20221698 [-] on Build GRCh38 62
          GenBankmRNA BF939615.1 - 1 - 2 Chr. 7 - 20219621 - 20221697 [-] on Build GRCh38 62
          GenBankmRNA AI350178.1 - 1 - 2 Chr. 7 - 20219697 - 20221696 [-] on Build GRCh38 62
          GenBankmRNA BF939614.1 - 1 - 2 Chr. 7 - 20219622 - 20221697 [-] on Build GRCh38 62
          GenBankmRNA AI401709.1 - 1 - 2 Chr. 7 - 20219666 - 20221700 [-] on Build GRCh38 62
          GenBankmRNA BX093269.1 - 4 - 5 381 Chr. 7 - 20217594 - 20221448 [+] on Build GRCh38 62
          GenBankmRNA AI652854.1 - 1 - 2 Chr. 7 - 20219613 - 20221697 [-] on Build GRCh38 62
          GenBankmRNA BF110962.1 - 1 - 2 Chr. 7 - 20219611 - 20221704 [-] on Build GRCh38 62
          GenBankmRNA AI340059.1 - 1 - 2 Chr. 7 - 20219625 - 20221697 [-] on Build GRCh38 62
          GenBankmRNA BF223968.1 - 1 - 2 Chr. 7 - 20219603 - 20221700 [-] on Build GRCh38 62
          GenBankmRNA BF475743.1 - 1 - 2 Chr. 7 - 20219611 - 20221699 [-] on Build GRCh38 62
          GenBankmRNA AI887799.1 - 1 - 2 Chr. 7 - 20219645 - 20221696 [-] on Build GRCh38 62
          GenBankmRNA CA306881.1 - 1 - 2 Chr. 7 - 20218284 - 20221704 [-] on Build GRCh38 62
          GenBankmRNA AI675079.1 - 1 - 2 Chr. 7 - 20219638 - 20221704 [-] on Build GRCh38 62
          GenBankmRNA BF224145.1 - 1 - 2 Chr. 7 - 20219611 - 20221700 [-] on Build GRCh38 62
          GenBankmRNA AW469903.1 - 1 - 2 Chr. 7 - 20219714 - 20221702 [-] on Build GRCh38 62
          GenBankmRNA AW510833.1 - 1 - 2 Chr. 7 - 20219612 - 20221700 [-] on Build GRCh38 62
          GenBankmRNA AW204499.1 - 1 - 2 Chr. 7 - 20219717 - 20221704 [-] on Build GRCh38 62
          GenBankmRNA W63621.1 - 313 Chr. - - [-] on Build 62
          GenBankmRNA BE348251.1 - 1 - 2 Chr. 7 - 20219628 - 20221700 [-] on Build GRCh38 62
          GenBankmRNA FN136974.1 - 1 - 2 52 Chr. 7 - 20219704 - 20221383 [+] on Build GRCh38 62
          GenBankmRNA BE674235.1 - 1 - 2 Chr. 7 - 20219621 - 20221700 [-] on Build GRCh38 62
          GenBankmRNA BF061801.1 - 1 - 2 Chr. 7 - 20219659 - 20221697 [-] on Build GRCh38 62
          GenBankmRNA BU675921.1 - 1 - 2 Chr. 7 - 20218289 - 20221704 [-] on Build GRCh38 62
          GenBankmRNA AI690298.1 - 1 - 2 Chr. 7 - 20219613 - 20221696 [-] on Build GRCh38 62
          GenBankmRNA BE669565.1 - 1 - 2 Chr. 7 - 20219612 - 20221700 [-] on Build GRCh38 62
          GenBankmRNA AI694145.1 - 1 - 2 Chr. 7 - 20219621 - 20221696 [-] on Build GRCh38 62
          GenBankmRNA BF061870.1 - 1 - 2 Chr. 7 - 20219621 - 20221690 [-] on Build GRCh38 62
          GenBankmRNA BU659281.1 - 3 - 4 441 Chr. 7 - 20217798 - 20221409 [+] on Build GRCh38 62
          GenBankmRNA W88841.1 - 382 Chr. - - [-] on Build 62
          GenBankmRNA AI689846.1 - 1 - 2 Chr. 7 - 20219620 - 20221696 [-] on Build GRCh38 62
          GenBankmRNA BF475792.1 - 1 - 2 Chr. 7 - 20219594 - 20221699 [-] on Build GRCh38 62
          GenBankmRNA AI650931.1 - 1 - 2 Chr. 7 - 20219624 - 20221697 [-] on Build GRCh38 62
          GenBankmRNA BE671445.1 - 1 - 2 Chr. 7 - 20219697 - 20221704 [-] on Build GRCh38 62
          GenBankmRNA BF222732.1 - 1 - 2 Chr. 7 - 20219610 - 20221697 [-] on Build GRCh38 62
          GenBankmRNA AI628065.1 - 1 - 2 Chr. 7 - 20219660 - 20221695 [-] on Build GRCh38 62
          GenBankmRNA AA831669.1 - 1 - 2 Chr. 7 - 20219649 - 20221704 [-] on Build GRCh38 62
          GenBankmRNA AA995320.1 - 1 - 2 Chr. 7 - 20219709 - 20221704 [-] on Build GRCh38 62
          GenBankmRNA BE550002.1 - 1 - 2 Chr. 7 - 20219562 - 20221697 [-] on Build GRCh38 62
          GenBankmRNA AW779745.1 - 1 - 2 Chr. 7 - 20219622 - 20221697 [-] on Build GRCh38 62
          GenBankmRNA BG149192.1 - 1 - 2 Chr. 7 - 20219621 - 20221700 [-] on Build GRCh38 62
          GenBankmRNA AI741598.1 - 1 - 2 Chr. 7 - 20219612 - 20221700 [-] on Build GRCh38 62
          GenBankmRNA AW024179.1 - 1 - 2 Chr. 7 - 20219634 - 20221697 [-] on Build GRCh38 62
          GenBankmRNA CD366064.1 - 1 - 2 Chr. 7 - 20219439 - 20221704 [-] on Build GRCh38 62
          GenBankmRNA AI692316.1 - 1 - 2 Chr. 7 - 20219614 - 20221696 [-] on Build GRCh38 62
          GenBankmRNA AW779730.1 - 1 - 2 Chr. 7 - 20219636 - 20221697 [-] on Build GRCh38 62
          GenBankmRNA AW301151.1 - 1 - 2 Chr. 7 - 20219696 - 20221704 [-] on Build GRCh38 62
          GenBankmRNA AI553988.1 - 1 - 2 Chr. 7 - 20219664 - 20221700 [-] on Build GRCh38 62
          GenBankmRNA BE048880.1 - 1 - 2 Chr. 7 - 20219686 - 20221697 [-] on Build GRCh38 62
          GenBankmRNA BF432870.1 - 1 - 2 Chr. 7 - 20219614 - 20221700 [-] on Build GRCh38 62
          GenBankmRNA AI767974.1 - 1 - 2 Chr. 7 - 20219663 - 20221700 [-] on Build GRCh38 62
          GenBankmRNA BF594766.1 - 1 - 2 Chr. 7 - 20219715 - 20221697 [-] on Build GRCh38 62
          GenBankmRNA AI458622.1 - 1 - 2 Chr. 7 - 20219666 - 20221696 [-] on Build GRCh38 62
          GenBankmRNA BF224090.1 - 1 - 2 Chr. 7 - 20218307 - 20221700 [-] on Build GRCh38 62
          GenBankmRNA AA680103.1 - 1 - 2 Chr. 7 - 20219694 - 20221652 [-] on Build GRCh38 62

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream metastasis associated in colon cancer 1 MACC1 7A5,
          SH3BP4L
          -2.2kb Anti-sense Chr. 7 - 20134655 - 20217390 on NCBI Build GRCh38
          Downstream uncharacterized LOC101927769 LOC101927769 75.0kb Sense Chr. 7 - 20296708 - 20311712 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0700006841.hg.1 ENST00000430859.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006841.hg.1 ENST00000430859.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006841.hg.1 ENST00000430859.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006841.hg.1 ENST00000430859.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006841.hg.1 ENST00000430859.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006841.hg.1 ENST00000430859.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006841.hg.1 ENST00000430859.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006841.hg.1 ENST00000439058.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006841.hg.1 ENST00000439058.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006841.hg.1 ENST00000439058.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006841.hg.1 ENST00000439058.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006841.hg.1 ENST00000439058.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006841.hg.1 ENST00000439058.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006841.hg.1 ENST00000439058.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006841.hg.1 ENST00000439058.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006841.hg.1 ENST00000439058.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006841.hg.1 ENST00000439058.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006841.hg.1 ENST00000439058.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006841.hg.1 ENST00000439058.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006841.hg.1 ENST00000439446.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006841.hg.1 ENST00000439446.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006841.hg.1 ENST00000439446.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006841.hg.1 ENST00000439446.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006841.hg.1 ENST00000439446.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006841.hg.1 ENST00000439446.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006841.hg.1 ENST00000439446.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006841.hg.1 ENST00000430859.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006841.hg.1 ENST00000430859.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006841.hg.1 ENST00000430859.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006841.hg.1 ENST00000430859.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006841.hg.1 ENST00000430859.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006841.hg.1 ENST00000430859.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006841.hg.1 ENST00000430859.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006841.hg.1 ENST00000439058.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006841.hg.1 ENST00000439058.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006841.hg.1 ENST00000439058.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006841.hg.1 ENST00000439058.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006841.hg.1 ENST00000439058.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006841.hg.1 ENST00000439058.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006841.hg.1 ENST00000439058.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006841.hg.1 ENST00000439058.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006841.hg.1 ENST00000439058.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006841.hg.1 ENST00000439058.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006841.hg.1 ENST00000439058.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006841.hg.1 ENST00000439058.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006841.hg.1 ENST00000439446.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006841.hg.1 ENST00000439446.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006841.hg.1 ENST00000439446.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006841.hg.1 ENST00000439446.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006841.hg.1 ENST00000439446.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006841.hg.1 ENST00000439446.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006841.hg.1 ENST00000439446.1 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-ffb8c786-92mpb:80/100.66.129.117:80.
          git-commit: 1a801765356e65be0b6ed8595ed79b52bb698fed
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0