Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929161 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_134641.1 | 2 - 3 | 343 | 122 | ||
GenBank mRNA | AA885339.1 | - | 2 - 3 | 122 | ||
BX099416.1 | - | 2 - 3 | 343 | 122 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | anaphase promoting complex subunit 4 | ANAPC4 | APC4 | -116.7kb | Anti-sense | Chr. 4 - 25377226 - 25418498 on NCBI Build GRCh38 |
Downstream | solute carrier family 34 member 2 | SLC34A2 |
NAPI-3B, NAPI-IIb, NPTIIb |
100.8kb | Anti-sense | Chr. 4 - 25655813 - 25678748 on NCBI Build GRCh38 |
Set Membership: |
![]() ![]() ![]() |