Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011988.hg.1 |
ENST00000623792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011988.hg.1 |
ENST00000626304.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011988.hg.1 |
ENST00000626304.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011988.hg.1 |
ENST00000626304.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011988.hg.1 |
ENST00000626304.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011988.hg.1 |
ENST00000626304.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011988.hg.1 |
ENST00000626304.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011988.hg.1 |
ENST00000628780.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011988.hg.1 |
ENST00000628780.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011988.hg.1 |
ENST00000628780.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011988.hg.1 |
ENST00000628780.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011988.hg.1 |
ENST00000628780.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011988.hg.1 |
ENST00000628780.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011988.hg.1 |
ENST00000628780.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011988.hg.1 |
ENST00000628780.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011988.hg.1 |
ENST00000628780.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011988.hg.1 |
ENST00000628780.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011988.hg.1 |
ENST00000628780.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011988.hg.1 |
ENST00000628780.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011988.hg.1 |
ENST00000628780.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011988.hg.1 |
ENST00000628780.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011988.hg.1 |
ENST00000623792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011988.hg.1 |
ENST00000626304.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011988.hg.1 |
ENST00000626304.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011988.hg.1 |
ENST00000626304.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011988.hg.1 |
ENST00000626304.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011988.hg.1 |
ENST00000626304.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011988.hg.1 |
ENST00000626304.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011988.hg.1 |
ENST00000628780.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011988.hg.1 |
ENST00000628780.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011988.hg.1 |
ENST00000628780.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011988.hg.1 |
ENST00000628780.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011988.hg.1 |
ENST00000628780.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011988.hg.1 |
ENST00000628780.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011988.hg.1 |
ENST00000628780.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011988.hg.1 |
ENST00000628780.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011988.hg.1 |
ENST00000628780.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011988.hg.1 |
ENST00000628780.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011988.hg.1 |
ENST00000628780.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011988.hg.1 |
ENST00000628780.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011988.hg.1 |
ENST00000628780.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011988.hg.1 |
ENST00000628780.1 |
Ensembl |