Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011828.hg.1 |
ENST00000563583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011828.hg.1 |
ENST00000563583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011828.hg.1 |
ENST00000563583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011828.hg.1 |
ENST00000564292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011828.hg.1 |
ENST00000564292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011828.hg.1 |
ENST00000564292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011828.hg.1 |
ENST00000564292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011828.hg.1 |
ENST00000564292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011828.hg.1 |
ENST00000564292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011828.hg.1 |
ENST00000564292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011828.hg.1 |
ENST00000564292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011828.hg.1 |
ENST00000585902.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011828.hg.1 |
ENST00000585902.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011828.hg.1 |
ENST00000585902.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011828.hg.1 |
ENST00000625040.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011828.hg.1 |
ENST00000625040.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011828.hg.1 |
ENST00000625040.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011828.hg.1 |
ENST00000625040.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011828.hg.1 |
ENST00000563583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011828.hg.1 |
ENST00000563583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011828.hg.1 |
ENST00000563583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011828.hg.1 |
ENST00000564292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011828.hg.1 |
ENST00000564292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011828.hg.1 |
ENST00000564292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011828.hg.1 |
ENST00000564292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011828.hg.1 |
ENST00000564292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011828.hg.1 |
ENST00000564292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011828.hg.1 |
ENST00000564292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011828.hg.1 |
ENST00000564292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011828.hg.1 |
ENST00000585902.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011828.hg.1 |
ENST00000585902.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011828.hg.1 |
ENST00000585902.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011828.hg.1 |
ENST00000625040.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011828.hg.1 |
ENST00000625040.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011828.hg.1 |
ENST00000625040.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011828.hg.1 |
ENST00000625040.1 |
Ensembl |