Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04939918_g1
          See other TBC1D25 GE Assays ›
          Gene Symbol:
          TBC1D25
          Gene Name:
          TBC1 domain family member 25
          Gene Aliases:
          MG81, OATL1
          Chromosome Location:
          Chr.X: 48539687 - 48562609 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04939918_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TBC1D25
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002536.2 NP_002527.1 5 - 6 815 93
          GenBank mRNA AB449912.1 - 5 - 6 704 93
          AK293521.1 - 4 - 5 645 93
          AK293875.1 - 5 - 6 774 93
          AK294776.1 - 5 - 6 713 93
          AK297924.1 - 2 - 3 220 93
          AK298884.1 - 4 - 5 674 93
          AK316199.1 - 3 - 4 495 93
          BC036942.1 - 2 - 3 244 93
          BC052609.1 - 4 - 5 605 93
          BC082255.1 - 3 - 4 715 93
          BC101817.1 - 5 - 6 741 8069323 93
          BC101819.1 - 5 - 6 741 8069325 93
          BC125088.1 - 5 - 6 730 40117866 93
          BC125089.1 - 4 - 5 575 93
          L08240.1 - 4 - 5 594 93
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DC309515.1 - 3 - 4 486 Chr. X - 48539729 - 48559707 [+] on Build GRCh38 93
          GenBankmRNA BQ646486.1 - 3 - 4 467 Chr. X - 48539770 - 48559988 [+] on Build GRCh38 93
          GenBankmRNA BX329059.1 - 1 - 2 Chr. X - 48559168 - 48560358 [-] on Build GRCh38 93
          GenBankmRNA BQ644832.1 - 3 - 4 467 Chr. X - 48539770 - 48559968 [+] on Build GRCh38 93
          GenBankmRNA HY151026.1 - 1 - 2 139 Chr. X - 48559207 - 48559924 [+] on Build GRCh38 93
          GenBankmRNA AL550349.3 - 3 - 4 380 Chr. X - 48544960 - 48560204 [+] on Build GRCh38 93
          GenBankmRNA AA293451.1 - 3 - 4 339 Chr. X - 48539897 - 48559823 [+] on Build GRCh38 92
          GenBankmRNA AL048477.2 - 1 - 2 195 Chr. X - 48559156 - 48560168 [+] on Build GRCh38 93
          GenBankmRNA DC296843.1 - 3 - 4 379 Chr. X - 48541380 - 48559790 [+] on Build GRCh38 93
          GenBankmRNA BM700569.1 - 2 - 3 274 Chr. X - 48558939 - 48559929 [+] on Build GRCh38 93
          GenBankmRNA DC338035.1 - 4 - 5 488 Chr. X - 48539860 - 48559685 [+] on Build GRCh38 93
          GenBankmRNA DA031057.1 - 3 - 4 509 Chr. X - 48539729 - 48559669 [+] on Build GRCh38 93
          GenBankmRNA AL036540.1 - 1 - 2 108 Chr. X - 48559288 - 48560010 [+] on Build GRCh38 93
          GenBankmRNA BQ651229.1 - 3 - 4 467 Chr. X - 48539770 - 48560041 [+] on Build GRCh38 93
          GenBankmRNA DC357458.1 - 2 - 3 220 Chr. X - 48558993 - 48559958 [+] on Build GRCh38 93
          GenBankmRNA DC344194.1 - 2 - 3 254 Chr. X - 48558959 - 48559912 [+] on Build GRCh38 93
          GenBankmRNA DA012386.1 - 3 - 4 505 Chr. X - 48539732 - 48559662 [+] on Build GRCh38 93
          GenBankmRNA HY128283.1 - 2 - 3 231 Chr. X - 48558992 - 48559884 [+] on Build GRCh38 93
          GenBankmRNA DC386828.1 - 4 - 5 487 Chr. X - 48539860 - 48559683 [+] on Build GRCh38 93
          GenBankmRNA DC407937.1 - 2 - 3 221 Chr. X - 48558993 - 48559923 [+] on Build GRCh38 93
          GenBankmRNA BG825374.1 - 4 - 5 610 Chr. X - 48539742 - 48559886 [+] on Build GRCh38 93
          GenBankmRNA DC315157.1 - 4 - 5 452 Chr. X - 48539895 - 48559728 [+] on Build GRCh38 93
          GenBankmRNA BF355438.1 - 366 Chr. - - [-] on Build 93
          GenBankmRNA DN999697.1 - 4 - 5 598 Chr. X - 48539755 - 48559691 [+] on Build GRCh38 93
          GenBankmRNA CB146082.1 - 3 - 4 500 Chr. X - 48539737 - 48559685 [+] on Build GRCh38 93
          GenBankmRNA AJ973530.1 - 1 - 2 Chr. X - 48559297 - 48560123 [-] on Build GRCh38 93
          GenBankmRNA DC326625.1 - 2 - 3 220 Chr. X - 48558993 - 48559962 [+] on Build GRCh38 93
          GenBankmRNA DA598712.1 - 3 - 4 496 Chr. X - 48539742 - 48559666 [+] on Build GRCh38 93

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream emopamil binding protein (sterol isomerase) EBP CDPX2,
          CHO2,
          CPX,
          CPXD,
          MEND
          -30.4kb Sense Chr. X - 48521776 - 48528716 on NCBI Build GRCh38
          Downstream RNA binding motif (RNP1,
          RRM) protein 3
          RBM3 IS1-RNPL,
          RNPL
          11.7kb Sense Chr. X - 48574353 - 48581165 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK293521.1, AK297924.1, AK316199.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TBC1D25

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          autophagy
          regulation of vesicle fusion
          activation of GTPase activity
          regulation of autophagosome maturation
          GTPase activator activity
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000418627.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000418627.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000418627.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000418627.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000418627.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000418627.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000476141.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000476141.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000476141.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000476141.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000476141.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000476141.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000476141.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000476141.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000476141.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000481090.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000481090.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000481090.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000481090.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000481090.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000481090.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000481090.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000481090.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000481090.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000494495.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000494495.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000494495.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000494495.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000494495.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 ENST00000494495.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000376771.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000418627.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000418627.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000418627.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000418627.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000418627.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000418627.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000476141.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000476141.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000476141.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000476141.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000476141.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000476141.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000476141.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000476141.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000476141.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000481090.6 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000481090.6 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000481090.6 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000481090.6 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000481090.6 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000481090.6 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000481090.6 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000481090.6 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000481090.6 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000494495.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000494495.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000494495.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000494495.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000494495.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 ENST00000494495.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 NM_002536.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00007194.hg.1 NM_002536.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0