Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04941464_m1
          See other APOBEC3G GE Assays ›
          Gene Symbol:
          APOBEC3G
          Gene Name:
          apolipoprotein B mRNA editing enzyme catalytic subunit 3G
          Gene Aliases:
          A3G, ARCD, ARP-9, ARP9, CEM-15, CEM15, MDS019, bK150C2.7, dJ494G10.1
          Chromosome Location:
          Chr.22: 39077005 - 39087743 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04941464_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: APOBEC3G
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006724290.1 XP_006724353.1 1 - 2 301 70
          XM_017028904.1 XP_016884393.1 1 - 2 301 70
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA HY076848.1 - 1 - 2 312 Chr. 22 - 39077068 - 39081095 [+] on Build GRCh38 70
          GenBankmRNA DB008551.1 - 1 - 2 306 Chr. 22 - 39077079 - 39081201 [+] on Build GRCh38 70
          GenBankmRNA BU598517.1 - 1 - 2 300 Chr. 22 - 39077083 - 39081566 [+] on Build GRCh38 70
          GenBankmRNA HY075131.1 - 1 - 2 312 Chr. 22 - 39077068 - 39081104 [+] on Build GRCh38 70
          GenBankmRNA DA925597.1 - 1 - 2 61 Chr. 22 - 39040918 - 39081687 [+] on Build GRCh38 70
          GenBankmRNA DB443624.1 - 1 - 2 105 Chr. 22 - 39077275 - 39081520 [+] on Build GRCh38 70

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream apolipoprotein B mRNA editing enzyme catalytic subunit 3F APOBEC3F A3F,
          ARP8,
          BK150C2.4.MRNA,
          KA6
          -21.1kb Sense Chr. 22 - 39020454 - 39055972 on NCBI Build GRCh38
          Downstream apolipoprotein B mRNA editing enzyme catalytic subunit 3H APOBEC3H A3H,
          ARP-10,
          ARP10
          16.0kb Sense Chr. 22 - 39097204 - 39104067 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the XM_006724290.1, XM_017028904.1 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of defense response to virus by host
          cytidine deamination
          negative regulation of transposition
          base conversion or substitution editing
          negative regulation of viral genome replication
          innate immune response
          negative regulation of single stranded viral RNA replication via double stranded DNA intermediate
          negative regulation of viral process
          defense response to virus
          DNA cytosine deamination
          RNA binding
          cytidine deaminase activity
          protein binding
          zinc ion binding
          dCTP deaminase activity
          protein homodimerization activity
          deoxycytidine deaminase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000407997.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000407997.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000407997.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000407997.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000407997.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000407997.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000407997.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000407997.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000407997.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000407997.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000407997.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000407997.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000461827.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000461827.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000461827.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000461827.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000461827.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000461827.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000463934.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000463934.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000463934.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000463934.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000463934.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000463934.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000480000.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000480000.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000480000.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000480000.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000480000.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000480000.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000480000.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000481958.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000481958.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000481958.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000481958.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000481958.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000481958.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000481958.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000486357.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000486357.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000486357.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000494150.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000494150.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000494150.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000494150.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000494150.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000494150.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000494150.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 ENST00000494150.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 NM_021822.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 NM_021822.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 NM_021822.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 NM_021822.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 NM_021822.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 NM_021822.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 NM_021822.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 NM_021822.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 NM_021822.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 NM_021822.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 NM_021822.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009274.hg.1 NM_021822.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000407997.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000407997.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000407997.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000407997.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000407997.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000407997.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000407997.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000407997.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000407997.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000407997.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000407997.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000407997.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000461827.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000461827.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000461827.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000461827.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000461827.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000461827.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000463934.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000463934.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000463934.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000463934.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000463934.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000463934.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000480000.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000480000.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000480000.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000480000.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000480000.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000480000.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000480000.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000481958.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000481958.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000481958.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000481958.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000481958.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000481958.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000481958.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000486357.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000486357.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000486357.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000494150.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000494150.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000494150.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000494150.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000494150.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000494150.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000494150.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 ENST00000494150.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 NM_021822.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 NM_021822.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 NM_021822.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 NM_021822.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 NM_021822.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 NM_021822.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 NM_021822.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 NM_021822.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 NM_021822.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 NM_021822.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 NM_021822.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009274.hg.1 NM_021822.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0