Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04941695_g1
          See other POU5F1P4 GE Assays ›
          Gene Symbol:
          POU5F1P4
          Gene Name:
          POU class 5 homeobox 1 pseudogene 4
          Gene Aliases:
          Oct4-pg4, POU5FLC1
          Chromosome Location:
          Chr.1: 155433180 - 155434262 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04941695_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: POU5F1P4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_034180.1 1 414 78
          GenBank mRNA AF268613.1 - 1 414 78
          DQ851566.1 - 1 414 78
          FM178001.1 - 1 292 78
          FM178003.1 - 1 292 78
          FM178004.1 - 1 292 78
          FM178009.1 - 1 292 78
          GU480814.1 - 1 415 78
          GU480831.1 - 1 416 78
          GU480834.1 - 1 415 78
          GU480836.1 - 1 415 78
          GU480839.1 - 1 415 78
          GU480848.1 - 1 415 78
          GU480853.1 - 1 416 78
          GU480854.1 - 1 415 78
          GU480855.1 - 1 413 78
          GU480857.1 - 1 415 78
          GU480858.1 - 1 415 78
          GU480859.1 - 1 415 78
          GU480860.1 - 1 414 78
          GU480861.1 - 1 416 78
          GU480862.1 - 1 415 78
          GU480863.1 - 1 415 78
          GU480865.1 - 1 416 78
          GU480866.1 - 1 415 78
          GU480867.1 - 1 415 78
          GU480869.1 - 1 415 78
          GU480870.1 - 1 415 78
          GU480871.1 - 1 415 78
          GU480872.1 - 1 415 78
          GU480873.1 - 1 415 78
          GU480874.1 - 1 415 78
          GU480875.1 - 1 415 78
          GU480879.1 - 1 415 78
          GU480881.1 - 1 415 78
          GU480882.1 - 1 415 78
          GU480890.1 - 1 414 78
          GU480894.1 - 1 415 78
          GU480895.1 - 1 415 78
          GU480898.1 - 1 414 78
          GU480900.1 - 1 414 78
          GU480904.1 - 1 415 78
          GU480914.1 - 1 414 78
          HQ122676.1 - 1 415 78
          HQ122677.1 - 1 415 78
          HQ122678.1 - 1 413 78
          HQ122679.1 - 1 77
          HQ122680.1 - 1 415 78
          HQ122681.1 - 1 415 78
          HQ122682.1 - 1 415 78
          HQ122683.1 - 1 416 78
          HQ122684.1 - 1 78
          HQ122685.1 - 1 413 78
          HQ122686.1 - 1 415 78
          HQ122687.1 - 1 415 78
          HQ122688.1 - 1 414 78
          HQ122689.1 - 1 413 78
          HQ122690.1 - 1 415 78
          HQ122695.1 - 1 414 78
          HQ122696.1 - 1 414 78
          HQ122697.1 - 1 78
          HQ122698.1 - 1 415 78
          HQ122699.1 - 1 415 78
          HQ122700.1 - 1 415 78
          HQ122702.1 - 1 415 78
          HQ122703.1 - 1 415 78
          HQ122705.1 - 1 415 78
          HQ122706.1 - 1 415 78
          HQ122707.1 - 1 414 78
          HQ122708.1 - 1 415 78
          HQ122731.1 - 1 415 78
          HQ122732.1 - 1 415 78
          HQ122733.1 - 1 415 78
          HQ122734.1 - 1 415 78
          HQ122735.1 - 1 415 78
          HQ122736.1 - 1 415 78
          HQ122740.1 - 1 415 78
          HQ122741.1 - 1 415 78
          HQ122742.1 - 1 415 78
          HQ122743.1 - 1 415 78
          HQ122744.1 - 1 415 78
          HQ122745.1 - 1 415 78
          HQ122746.1 - 1 415 78
          HQ122747.1 - 1 415 78
          HQ122748.1 - 1 415 78
          HQ122757.1 - 1 415 78
          HQ122758.1 - 1 415 78
          HQ122760.1 - 1 415 78
          HQ122761.1 - 1 415 78
          HQ122762.1 - 1 415 78
          HQ122763.1 - 1 415 78
          HQ122764.1 - 1 415 78
          HQ122765.1 - 1 415 78
          HQ122766.1 - 1 415 78
          HQ122767.1 - 1 415 78
          HQ122768.1 - 1 415 78
          HQ122769.1 - 1 414 78
          HQ122771.1 - 1 415 78
          HQ122772.1 - 1 415 78
          HQ122774.1 - 1 415 78
          HQ122775.1 - 1 415 78
          HQ122777.1 - 1 414 78
          HQ122778.1 - 1 415 78
          HQ122779.1 - 1 415 78
          HQ122780.1 - 1 415 78
          HQ122798.1 - 1 415 78
          HQ122799.1 - 1 414 78
          HQ122800.1 - 1 416 78
          HQ122801.1 - 1 415 78
          HQ122802.1 - 1 413 78
          HQ122803.1 - 1 78
          HQ122804.1 - 1 415 78
          HQ122805.1 - 1 415 78
          HQ122806.1 - 1 415 78
          HQ122807.1 - 1 415 78
          HQ122808.1 - 1 415 78
          HQ122809.1 - 1 413 78
          HQ122810.1 - 1 415 78
          HQ122811.1 - 1 415 78
          HQ122812.1 - 1 415 78
          HQ122813.1 - 1 415 78
          HQ122814.1 - 1 413 78
          HQ122815.1 - 1 415 78
          HQ122832.1 - 1 415 78
          HQ122833.1 - 1 415 78
          HQ122834.1 - 1 415 78
          HQ122835.1 - 1 413 78
          HQ122836.1 - 1 78
          HQ122837.1 - 1 415 78
          HQ122838.1 - 1 413 78
          HQ122839.1 - 1 415 78
          HQ122840.1 - 1 78
          HQ122858.1 - 1 415 78
          HQ122888.1 - 1 415 78
          HQ122889.1 - 1 415 78
          HQ122890.1 - 1 415 78
          HQ122904.1 - 1 415 78
          HQ122906.1 - 1 79
          HQ122908.1 - 1 415 78
          HQ122914.1 - 1 415 78

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline