Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04961755_gH
          See other MIR548AJ2 GE Assays ›
          Gene Symbol:
          MIR548AJ2
          Gene Name:
          microRNA 548aj-2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 38023895 - 38023986 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04961755_gH
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MIR548AJ2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_039674.1 1 42 91

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream synaptotagmin like 5 SYTL5 slp5 Anti-sense Chr. X - 37888915 - 38128820 on NCBI Build GRCh38
          Downstream sushi repeat containing protein,
          X-linked
          SRPX DRS,
          ETX1,
          HEL-S-83p,
          SRPX1
          125.3kb Sense Chr. X - 38149335 - 38220924 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0200012563.hg.1 ENST00000401149.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008702.hg.1 ENST00000583581.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008435.hg.1 ENST00000408077.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006492.hg.1 ENST00000618214.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011102.hg.1 ENST00000516922.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007527.hg.1 ENST00000617507.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009385.hg.1 ENST00000408539.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009385.hg.1 ENST00000408539.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009385.hg.1 NR_039674.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009385.hg.1 NR_039674.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011436.hg.1 ENST00000584743.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012397.hg.1 ENST00000585282.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009413.hg.1 ENST00000408523.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006850.hg.1 ENST00000408071.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006849.hg.1 ENST00000577673.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015198.hg.1 ENST00000516195.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014307.hg.1 ENST00000411375.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007834.hg.1 ENST00000408489.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009322.hg.1 ENST00000582731.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007301.hg.1 ENST00000583324.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009181.hg.1 ENST00000581952.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008035.hg.1 ENST00000410229.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010223.hg.1 ENST00000580555.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010183.hg.1 ENST00000615001.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008456.hg.1 ENST00000408664.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008298.hg.1 ENST00000516644.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010979.hg.1 ENST00000620698.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007505.hg.1 ENST00000577415.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008001.hg.1 ENST00000390764.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011778.hg.1 ENST00000584487.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400008107.hg.1 ENST00000408152.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008812.hg.1 ENST00000579683.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008812.hg.1 ENST00000579683.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300008657.hg.1 ENST00000581281.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008987.hg.1 ENST00000408540.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008957.hg.1 ENST00000408271.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014541.hg.1 ENST00000408362.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008446.hg.1 ENST00000585220.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015198.hg.1 ENST00000516195.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500008987.hg.1 ENST00000408540.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009385.hg.1 ENST00000408539.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009385.hg.1 ENST00000408539.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009385.hg.1 NR_039674.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00009385.hg.1 NR_039674.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00009322.hg.1 ENST00000582731.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010223.hg.1 ENST00000580555.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600008446.hg.1 ENST00000585220.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008702.hg.1 ENST00000583581.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006492.hg.1 ENST00000618214.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600007301.hg.1 ENST00000583324.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008812.hg.1 ENST00000579683.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008812.hg.1 ENST00000579683.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400008107.hg.1 ENST00000408152.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300009181.hg.1 ENST00000581952.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008035.hg.1 ENST00000410229.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007505.hg.1 ENST00000577415.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012397.hg.1 ENST00000585282.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010183.hg.1 ENST00000615001.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012563.hg.1 ENST00000401149.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100011102.hg.1 ENST00000516922.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100014307.hg.1 ENST00000411375.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900009413.hg.1 ENST00000408523.3 Ensembl
          Affymetrix Human Clariom D Assay TC1300006850.hg.1 ENST00000408071.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200011778.hg.1 ENST00000584487.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006849.hg.1 ENST00000577673.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900010979.hg.1 ENST00000620698.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200011436.hg.1 ENST00000584743.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100008001.hg.1 ENST00000390764.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200014541.hg.1 ENST00000408362.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100008456.hg.1 ENST00000408664.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008435.hg.1 ENST00000408077.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400007527.hg.1 ENST00000617507.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008298.hg.1 ENST00000516644.2 Ensembl
          Affymetrix Human Clariom D Assay TC1300008957.hg.1 ENST00000408271.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007834.hg.1 ENST00000408489.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300008657.hg.1 ENST00000581281.1 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0