Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008856.hg.1 |
ENST00000332349.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008856.hg.1 |
ENST00000332349.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008856.hg.1 |
ENST00000332349.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008856.hg.1 |
ENST00000332349.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008856.hg.1 |
ENST00000332349.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008856.hg.1 |
ENST00000546177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008856.hg.1 |
ENST00000546177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008856.hg.1 |
ENST00000546177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008856.hg.1 |
NM_001261833.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008856.hg.1 |
NM_001261833.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008856.hg.1 |
NM_001261833.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008856.hg.1 |
NM_001261833.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008856.hg.1 |
NM_001261833.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008856.hg.1 |
NM_001261833.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008856.hg.1 |
NM_003772.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008856.hg.1 |
NM_003772.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008856.hg.1 |
NM_003772.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008856.hg.1 |
NM_003772.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008856.hg.1 |
NM_003772.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008856.hg.1 |
NM_003772.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008856.hg.1 |
NM_003772.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008856.hg.1 |
ENST00000332349.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008856.hg.1 |
ENST00000332349.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008856.hg.1 |
ENST00000332349.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008856.hg.1 |
ENST00000332349.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008856.hg.1 |
ENST00000332349.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008856.hg.1 |
ENST00000546177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008856.hg.1 |
ENST00000546177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008856.hg.1 |
ENST00000546177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008856.hg.1 |
NM_001261833.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008856.hg.1 |
NM_001261833.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008856.hg.1 |
NM_001261833.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008856.hg.1 |
NM_001261833.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008856.hg.1 |
NM_001261833.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008856.hg.1 |
NM_001261833.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008856.hg.1 |
NM_003772.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008856.hg.1 |
NM_003772.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008856.hg.1 |
NM_003772.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008856.hg.1 |
NM_003772.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008856.hg.1 |
NM_003772.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008856.hg.1 |
NM_003772.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008856.hg.1 |
NM_003772.3 |
Refseq |