Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04964166_s1
          See other PYDC1 GE Assays ›
          Gene Symbol:
          PYDC1
          Gene Name:
          pyrin domain containing 1
          Gene Aliases:
          ASC2, POP1, PYC1, cPOP1
          Chromosome Location:
          Chr.16: 31215962 - 31217093 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04964166_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PYDC1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_152901.3 NP_690865.1 1 90 55
          GenBank mRNA AF086332.1 - 1 90 55
          AF454669.1 - 1 79 55
          AF467809.1 - 1 85 55
          AK313418.1 - 1 356 55
          AY163727.1 - 1 90 55
          BC105033.1 - 1 76 8144036 55
          BC105035.1 - 1 76 8144038 55

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PYDC1

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of innate immune response
          positive regulation of defense response to virus by host
          myeloid dendritic cell activation involved in immune response
          positive regulation of antigen processing and presentation of peptide antigen via MHC class II
          negative regulation of protein kinase activity
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          inflammatory response
          response to bacterium
          regulation of autophagy
          negative regulation of tumor necrosis factor-mediated signaling pathway
          positive regulation of actin filament polymerization
          regulation of protein stability
          negative regulation of NF-kappaB transcription factor activity
          interleukin-1 beta production
          positive regulation of chemokine production
          positive regulation of interferon-gamma production
          positive regulation of interleukin-6 production
          tumor necrosis factor-mediated signaling pathway
          positive regulation of activated T cell proliferation
          regulation of GTPase activity
          macropinocytosis
          innate immune response
          regulation of inflammatory response
          positive regulation of phagocytosis
          positive regulation of ERK1 and ERK2 cascade
          activation of cysteine-type endopeptidase activity
          negative regulation of NLRP3 inflammasome complex assembly
          positive regulation of T cell migration
          protein binding
          protein dimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1600010012.hg.1 ENST00000302964.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010012.hg.1 ENST00000302964.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010012.hg.1 ENST00000302964.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010012.hg.1 ENST00000302964.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010012.hg.1 ENST00000302964.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010012.hg.1 ENST00000568383.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010012.hg.1 ENST00000568383.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010012.hg.1 ENST00000568383.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010012.hg.1 NM_152901.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010012.hg.1 NM_152901.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010012.hg.1 NM_152901.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010012.hg.1 NM_152901.3 Refseq
          Affymetrix Human Clariom D Assay TC1600010012.hg.1 ENST00000302964.3 Ensembl
          Affymetrix Human Clariom D Assay TC1600010012.hg.1 ENST00000302964.3 Ensembl
          Affymetrix Human Clariom D Assay TC1600010012.hg.1 ENST00000302964.3 Ensembl
          Affymetrix Human Clariom D Assay TC1600010012.hg.1 ENST00000302964.3 Ensembl
          Affymetrix Human Clariom D Assay TC1600010012.hg.1 ENST00000302964.3 Ensembl
          Affymetrix Human Clariom D Assay TC1600010012.hg.1 ENST00000568383.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600010012.hg.1 ENST00000568383.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600010012.hg.1 ENST00000568383.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600010012.hg.1 NM_152901.3 Refseq
          Affymetrix Human Clariom D Assay TC1600010012.hg.1 NM_152901.3 Refseq
          Affymetrix Human Clariom D Assay TC1600010012.hg.1 NM_152901.3 Refseq
          Affymetrix Human Clariom D Assay TC1600010012.hg.1 NM_152901.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0