Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012367.hg.1 |
ENST00000422013.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012367.hg.1 |
ENST00000422013.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012367.hg.1 |
ENST00000422013.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012367.hg.1 |
ENST00000422013.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012367.hg.1 |
ENST00000458490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012367.hg.1 |
ENST00000458490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012367.hg.1 |
ENST00000458490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012367.hg.1 |
NR_110584.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012367.hg.1 |
NR_110584.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012367.hg.1 |
NR_110584.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012367.hg.1 |
NR_110584.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012367.hg.1 |
NR_110584.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012367.hg.1 |
ENST00000422013.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012367.hg.1 |
ENST00000422013.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012367.hg.1 |
ENST00000422013.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012367.hg.1 |
ENST00000422013.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012367.hg.1 |
ENST00000458490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012367.hg.1 |
ENST00000458490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012367.hg.1 |
ENST00000458490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012367.hg.1 |
NR_110584.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012367.hg.1 |
NR_110584.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012367.hg.1 |
NR_110584.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012367.hg.1 |
NR_110584.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012367.hg.1 |
NR_110584.1 |
Refseq |