Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04969997_m1
          See other CCR2 GE Assays ›
          Gene Symbol:
          CCR2
          Gene Name:
          C-C motif chemokine receptor 2
          Gene Aliases:
          CC-CKR-2, CCR-2, CCR2A, CCR2B, CD192, CKR2, CKR2A, CKR2B, CMKBR2, MCP-1-R
          Chromosome Location:
          Chr.3: 46353744 - 46360940 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04969997_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CCR2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_011534069.1 XP_011532371.1 1 - 2 52 100

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the XM_011534069.1 transcripts.

          Set Membership:

          Amplicon length between 86 and 100 Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel remodeling
          dendritic cell chemotaxis
          inflammatory response to antigenic stimulus
          positive regulation of T-helper 1 type immune response
          negative regulation of type 2 immune response
          cellular calcium ion homeostasis
          chemotaxis
          inflammatory response
          immune response
          cellular defense response
          G protein-coupled receptor signaling pathway
          negative regulation of adenylate cyclase activity
          positive regulation of cytosolic calcium ion concentration
          receptor signaling pathway via JAK-STAT
          aging
          response to wounding
          response to ionizing radiation
          regulation of vascular endothelial growth factor production
          positive regulation of gene expression
          positive regulation of T cell chemotaxis
          negative regulation of angiogenesis
          cytokine-mediated signaling pathway
          cellular homeostasis
          negative regulation of cell migration
          negative regulation of axon extension
          forebrain development
          positive regulation of fever generation
          response to lipopolysaccharide
          positive regulation of interferon-gamma production
          positive regulation of interleukin-2 production
          positive regulation of interleukin-6 production
          T-helper 17 cell chemotaxis
          defense response to bacterium
          negative regulation of eosinophil degranulation
          fat cell differentiation
          positive regulation of neuron differentiation
          positive regulation of alpha-beta T cell proliferation
          positive regulation of inflammatory response
          positive regulation of T cell activation
          positive regulation of apoptotic process by virus
          chemokine-mediated signaling pathway
          neuron death
          positive regulation of monocyte chemotaxis
          positive regulation of cell migration involved in sprouting angiogenesis
          positive regulation of immune complex clearance by monocytes and macrophages
          negative regulation of neural precursor cell proliferation
          positive regulation of monocyte extravasation
          positive regulation of CD8-positive, alpha-beta T cell extravasation
          positive regulation of astrocyte chemotaxis
          positive regulation of hematopoietic stem cell migration
          chemokine receptor activity
          C-C chemokine receptor activity
          protein kinase binding
          CCR2 chemokine receptor binding
          protein homodimerization activity
          chemokine (C-C motif) ligand 5 binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000292301.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000292301.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000292301.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000292301.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000292301.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000292301.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000292301.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000292301.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000292301.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000292301.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000292301.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000400888.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000400888.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000400888.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000400888.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000400888.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000400888.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000400888.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000400888.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000421659.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000421659.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000421659.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000421659.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000445132.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000445132.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000445132.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000445132.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000445132.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000445132.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000445132.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000445132.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000445132.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000445132.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000445132.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000465202.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000465202.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000465202.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000465202.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 ENST00000465202.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 NM_001123041.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 NM_001123041.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 NM_001123041.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 NM_001123041.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 NM_001123041.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 NM_001123041.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 NM_001123041.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 NM_001123041.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 NM_001123041.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 NM_001123041.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 NM_001123041.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 NM_001123396.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 NM_001123396.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 NM_001123396.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 NM_001123396.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 NM_001123396.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 NM_001123396.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 NM_001123396.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 NM_001123396.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 NM_001123396.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007256.hg.1 NM_001123396.2 Refseq
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000292301.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000292301.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000292301.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000292301.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000292301.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000292301.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000292301.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000292301.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000292301.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000292301.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000292301.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000400888.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000400888.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000400888.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000400888.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000400888.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000400888.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000400888.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000400888.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000421659.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000421659.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000421659.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000421659.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000445132.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000445132.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000445132.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000445132.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000445132.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000445132.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000445132.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000445132.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000445132.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000445132.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000445132.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000465202.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000465202.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000465202.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000465202.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 ENST00000465202.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 NM_001123041.2 Refseq
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 NM_001123041.2 Refseq
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 NM_001123041.2 Refseq
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 NM_001123041.2 Refseq
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 NM_001123041.2 Refseq
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 NM_001123041.2 Refseq
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 NM_001123041.2 Refseq
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 NM_001123041.2 Refseq
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 NM_001123041.2 Refseq
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 NM_001123041.2 Refseq
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 NM_001123041.2 Refseq
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 NM_001123396.2 Refseq
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 NM_001123396.2 Refseq
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 NM_001123396.2 Refseq
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 NM_001123396.2 Refseq
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 NM_001123396.2 Refseq
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 NM_001123396.2 Refseq
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 NM_001123396.2 Refseq
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 NM_001123396.2 Refseq
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 NM_001123396.2 Refseq
          Affymetrix Human Clariom D Assay TC0300007256.hg.1 NM_001123396.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0