Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04972549_s1
          See other CCR5 GE Assays ›
          Gene Symbol:
          CCR5
          Gene Name:
          C-C motif chemokine receptor 5 (gene/pseudogene)
          Gene Aliases:
          CC-CKR-5, CCCKR5, CCR-5, CD195, CKR-5, CKR5, CMKBR5, IDDM22
          Chromosome Location:
          Chr.3: 46370142 - 46376206 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04972549_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CCR5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000579.3 NP_000570.1 3 600 138
          NM_001100168.1 NP_001093638.1 3 365 138
          GenBank mRNA AF011500.1 - 1 247 138
          AF011501.1 - 1 247 138
          AF011502.1 - 1 247 138
          AF011503.1 - 1 247 138
          AF011504.1 - 1 246 138
          AF011505.1 - 1 247 138
          AF011506.1 - 1 247 138
          AF011507.1 - 1 247 138
          AF011508.1 - 1 247 138
          AF011509.1 - 1 247 138
          AF011510.1 - 1 247 138
          AF011511.1 - 1 247 138
          AF011512.1 - 1 247 138
          AF011513.1 - 1 247 138
          AF011514.1 - 1 247 138
          AF011515.1 - 1 247 138
          AF011516.1 - 1 247 138
          AF011517.1 - 1 247 138
          AF011518.1 - 1 247 138
          AF011519.1 - 1 247 138
          AF011520.1 - 1 247 138
          AF011521.1 - 1 247 138
          AF011522.1 - 1 247 138
          AF011523.1 - 1 247 138
          AF011524.1 - 1 247 138
          AF011525.1 - 1 247 138
          AF011526.1 - 1 247 138
          AF011527.1 - 1 247 138
          AF011528.1 - 1 247 138
          AF011529.1 - 1 247 138
          AF011530.1 - 1 247 138
          AF011531.1 - 1 247 138
          AF011532.1 - 1 247 138
          AF011533.1 - 1 247 138
          AF011534.1 - 1 247 138
          AF011535.1 - 1 247 138
          AF011536.1 - 1 247 138
          AF011537.1 - 1 247 138
          AK292132.1 - 2 297 138
          AK314882.1 - 2 389 138
          BC038398.1 - 2 286 5186388 138
          DA818906.1 - 2 389 138
          U54994.1 - 2 297 138
          U57840.1 - 2 269 138

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CCR5

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          dendritic cell chemotaxis
          calcium ion transport
          chemotaxis
          inflammatory response
          immune response
          cellular defense response
          cell surface receptor signaling pathway
          G protein-coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          cell-cell signaling
          release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
          fusion of virus membrane with host plasma membrane
          calcium-mediated signaling
          signaling
          chemokine-mediated signaling pathway
          response to cholesterol
          cellular response to lipopolysaccharide
          virus receptor activity
          actin binding
          phosphatidylinositol phospholipase C activity
          chemokine receptor activity
          protein binding
          coreceptor activity
          C-C chemokine receptor activity
          C-C chemokine binding
          chemokine (C-C motif) ligand 5 binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0300007257.hg.1 ENST00000292303.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007257.hg.1 ENST00000292303.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007257.hg.1 ENST00000292303.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007257.hg.1 ENST00000292303.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007257.hg.1 ENST00000292303.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007257.hg.1 ENST00000292303.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007257.hg.1 ENST00000445772.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007257.hg.1 ENST00000445772.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007257.hg.1 ENST00000445772.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007257.hg.1 ENST00000445772.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007257.hg.1 NM_000579.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007257.hg.1 NM_000579.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007257.hg.1 NM_000579.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007257.hg.1 NM_000579.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007257.hg.1 NM_000579.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007257.hg.1 NM_000579.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007257.hg.1 NM_000579.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007257.hg.1 NM_000579.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007257.hg.1 NM_001100168.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007257.hg.1 NM_001100168.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007257.hg.1 NM_001100168.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007257.hg.1 NM_001100168.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007257.hg.1 NM_001100168.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007257.hg.1 NM_001100168.1 Refseq
          Affymetrix Human Clariom D Assay TC0300007257.hg.1 ENST00000292303.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007257.hg.1 ENST00000292303.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007257.hg.1 ENST00000292303.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007257.hg.1 ENST00000292303.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007257.hg.1 ENST00000292303.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007257.hg.1 ENST00000292303.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007257.hg.1 ENST00000445772.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007257.hg.1 ENST00000445772.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007257.hg.1 ENST00000445772.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007257.hg.1 ENST00000445772.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007257.hg.1 NM_000579.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007257.hg.1 NM_000579.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007257.hg.1 NM_000579.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007257.hg.1 NM_000579.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007257.hg.1 NM_000579.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007257.hg.1 NM_000579.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007257.hg.1 NM_000579.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007257.hg.1 NM_000579.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007257.hg.1 NM_001100168.1 Refseq
          Affymetrix Human Clariom D Assay TC0300007257.hg.1 NM_001100168.1 Refseq
          Affymetrix Human Clariom D Assay TC0300007257.hg.1 NM_001100168.1 Refseq
          Affymetrix Human Clariom D Assay TC0300007257.hg.1 NM_001100168.1 Refseq
          Affymetrix Human Clariom D Assay TC0300007257.hg.1 NM_001100168.1 Refseq
          Affymetrix Human Clariom D Assay TC0300007257.hg.1 NM_001100168.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0