Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000511687.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000511687.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000511687.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000511687.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000511687.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000511687.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |