Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04978545_g1
          See other FGF10 GE Assays ›
          Gene Symbol:
          FGF10
          Gene Name:
          fibroblast growth factor 10
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 44301655 - 44389780 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04978545_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FGF10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_005248264.3 XP_005248321.1 1 - 2 625 109

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the XM_005248264.3 transcripts.

          Set Membership:

          Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          establishment of mitotic spindle orientation
          MAPK cascade
          angiogenesis
          metanephros development
          organ induction
          mesonephros development
          blood vessel remodeling
          metanephros morphogenesis
          determination of left/right symmetry
          salivary gland development
          negative regulation of cell population proliferation
          fibroblast growth factor receptor signaling pathway
          regulation of smoothened signaling pathway
          embryonic pattern specification
          epithelial cell migration
          positive regulation of epithelial cell migration
          positive regulation of keratinocyte proliferation
          regulation of phosphatidylinositol 3-kinase signaling
          peptidyl-tyrosine phosphorylation
          pituitary gland development
          embryonic genitalia morphogenesis
          thyroid gland development
          otic vesicle formation
          positive regulation of vascular endothelial growth factor receptor signaling pathway
          pancreas development
          hair follicle morphogenesis
          embryonic camera-type eye development
          actin cytoskeleton reorganization
          response to estradiol
          response to lipopolysaccharide
          positive regulation of ATPase activity
          lacrimal gland development
          regulation of activin receptor signaling pathway
          protein localization to cell surface
          somatic stem cell population maintenance
          phosphatidylinositol-3-phosphate biosynthetic process
          wound healing
          tissue regeneration
          odontogenesis of dentin-containing tooth
          muscle cell fate commitment
          positive regulation of MAPK cascade
          positive regulation of GTPase activity
          keratinocyte proliferation
          negative regulation of cell differentiation
          positive regulation of DNA repair
          positive regulation of DNA replication
          positive regulation of Notch signaling pathway
          positive regulation of transcription, DNA-templated
          positive regulation of mitotic cell cycle
          positive regulation of transcription by RNA polymerase II
          positive regulation of Ras protein signal transduction
          phosphatidylinositol phosphate biosynthetic process
          regulation of saliva secretion
          phosphatidylinositol-mediated signaling
          positive regulation of fibroblast proliferation
          spleen development
          thymus development
          embryonic digestive tract morphogenesis
          branching morphogenesis of an epithelial tube
          female genitalia morphogenesis
          male genitalia morphogenesis
          positive regulation of lymphocyte proliferation
          epithelial cell proliferation
          urothelial cell proliferation
          positive regulation of urothelial cell proliferation
          positive regulation of epithelial cell proliferation
          positive regulation of peptidyl-tyrosine phosphorylation
          white fat cell differentiation
          positive chemotaxis
          induction of positive chemotaxis
          smooth muscle cell differentiation
          positive regulation of keratinocyte migration
          radial glial cell differentiation
          positive regulation of epithelial cell proliferation involved in wound healing
          limb bud formation
          lung epithelium development
          lung saccule development
          bronchiole morphogenesis
          bud outgrowth involved in lung branching
          bud elongation involved in lung branching
          mesenchymal-epithelial cell signaling involved in lung development
          type II pneumocyte differentiation
          prostatic bud formation
          fibroblast growth factor receptor signaling pathway involved in mammary gland specification
          mammary gland bud formation
          submandibular salivary gland formation
          epithelial cell proliferation involved in salivary gland morphogenesis
          regulation of branching involved in salivary gland morphogenesis by mesenchymal-epithelial signaling
          branch elongation involved in salivary gland morphogenesis
          semicircular canal fusion
          mesenchymal cell differentiation involved in lung development
          secretion by lung epithelial cell involved in lung growth
          lung proximal/distal axis specification
          tear secretion
          positive regulation of white fat cell proliferation
          ERK1 and ERK2 cascade
          positive regulation of ERK1 and ERK2 cascade
          Harderian gland development
          positive regulation of hair follicle cell proliferation
          positive regulation of canonical Wnt signaling pathway
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          protein tyrosine kinase activity
          fibroblast growth factor receptor binding
          type 2 fibroblast growth factor receptor binding
          protein binding
          growth factor activity
          heparin binding
          1-phosphatidylinositol-3-kinase activity
          chemoattractant activity
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0500010651.hg.1 ENST00000264664.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010651.hg.1 ENST00000264664.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010651.hg.1 ENST00000264664.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010651.hg.1 ENST00000264664.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010651.hg.1 ENST00000264664.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010651.hg.1 ENST00000264664.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010651.hg.1 ENST00000264664.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010651.hg.1 ENST00000513107.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010651.hg.1 ENST00000513107.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010651.hg.1 ENST00000513107.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010651.hg.1 ENST00000513107.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010651.hg.1 ENST00000513107.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010651.hg.1 NM_004465.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010651.hg.1 NM_004465.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010651.hg.1 NM_004465.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010651.hg.1 NM_004465.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007309.hg.1 ENST00000502457.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007309.hg.1 ENST00000502457.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007309.hg.1 ENST00000502457.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007309.hg.1 NR_108034.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007309.hg.1 NR_108034.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007309.hg.1 NR_108034.1 Refseq
          Affymetrix Human Clariom D Assay TC0500007309.hg.1 ENST00000502457.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500007309.hg.1 ENST00000502457.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500007309.hg.1 ENST00000502457.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500007309.hg.1 NR_108034.1 Refseq
          Affymetrix Human Clariom D Assay TC0500007309.hg.1 NR_108034.1 Refseq
          Affymetrix Human Clariom D Assay TC0500007309.hg.1 NR_108034.1 Refseq
          Affymetrix Human Clariom D Assay TC0500010651.hg.1 ENST00000264664.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010651.hg.1 ENST00000264664.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010651.hg.1 ENST00000264664.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010651.hg.1 ENST00000264664.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010651.hg.1 ENST00000264664.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010651.hg.1 ENST00000264664.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010651.hg.1 ENST00000264664.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010651.hg.1 ENST00000513107.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500010651.hg.1 ENST00000513107.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500010651.hg.1 ENST00000513107.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500010651.hg.1 ENST00000513107.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500010651.hg.1 ENST00000513107.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500010651.hg.1 NM_004465.1 Refseq
          Affymetrix Human Clariom D Assay TC0500010651.hg.1 NM_004465.1 Refseq
          Affymetrix Human Clariom D Assay TC0500010651.hg.1 NM_004465.1 Refseq
          Affymetrix Human Clariom D Assay TC0500010651.hg.1 NM_004465.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline