Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04982536_s1
          See other HCN4 GE Assays ›
          Gene Symbol:
          HCN4
          Gene Name:
          hyperpolarization activated cyclic nucleotide gated potassium channel 4
          Gene Aliases:
          SSS2
          Chromosome Location:
          Chr.15: 73319859 - 73369264 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04982536_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HCN4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005477.2 NP_005468.1 1 664 96
          GenBank mRNA AJ132429.1 - 1 236 96
          AJ238850.1 - 1 668 94
          DQ854814.1 - 1 314 94

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AJ132429.1, AJ238850.1, NM_005477.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene HCN4

          Panther Classification:

          Molecular Function -

          ion channel transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of heart rate
          regulation of membrane depolarization
          cation transport
          muscle contraction
          blood circulation
          regulation of ion transmembrane transport
          sodium ion transmembrane transport
          regulation of membrane potential
          regulation of cardiac muscle contraction
          regulation of postsynaptic membrane potential
          cellular response to cAMP
          cellular response to cGMP
          potassium ion transmembrane transport
          SA node cell action potential
          regulation of heart rate by cardiac conduction
          intracellular cAMP activated cation channel activity
          voltage-gated sodium channel activity
          voltage-gated potassium channel activity
          cAMP binding
          identical protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1500009981.hg.1 ENST00000261917.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009981.hg.1 ENST00000261917.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009981.hg.1 ENST00000261917.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009981.hg.1 ENST00000261917.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009981.hg.1 ENST00000261917.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009981.hg.1 ENST00000261917.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009981.hg.1 ENST00000261917.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009981.hg.1 ENST00000261917.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009981.hg.1 ENST00000261917.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009981.hg.1 ENST00000261917.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009981.hg.1 ENST00000261917.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009981.hg.1 NM_005477.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009981.hg.1 NM_005477.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009981.hg.1 NM_005477.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009981.hg.1 NM_005477.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009981.hg.1 NM_005477.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009981.hg.1 NM_005477.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009981.hg.1 NM_005477.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009981.hg.1 NM_005477.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009981.hg.1 NM_005477.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009981.hg.1 NM_005477.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009981.hg.1 NM_005477.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009981.hg.1 ENST00000261917.3 Ensembl
          Affymetrix Human Clariom D Assay TC1500009981.hg.1 ENST00000261917.3 Ensembl
          Affymetrix Human Clariom D Assay TC1500009981.hg.1 ENST00000261917.3 Ensembl
          Affymetrix Human Clariom D Assay TC1500009981.hg.1 ENST00000261917.3 Ensembl
          Affymetrix Human Clariom D Assay TC1500009981.hg.1 ENST00000261917.3 Ensembl
          Affymetrix Human Clariom D Assay TC1500009981.hg.1 ENST00000261917.3 Ensembl
          Affymetrix Human Clariom D Assay TC1500009981.hg.1 ENST00000261917.3 Ensembl
          Affymetrix Human Clariom D Assay TC1500009981.hg.1 ENST00000261917.3 Ensembl
          Affymetrix Human Clariom D Assay TC1500009981.hg.1 ENST00000261917.3 Ensembl
          Affymetrix Human Clariom D Assay TC1500009981.hg.1 ENST00000261917.3 Ensembl
          Affymetrix Human Clariom D Assay TC1500009981.hg.1 ENST00000261917.3 Ensembl
          Affymetrix Human Clariom D Assay TC1500009981.hg.1 NM_005477.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009981.hg.1 NM_005477.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009981.hg.1 NM_005477.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009981.hg.1 NM_005477.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009981.hg.1 NM_005477.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009981.hg.1 NM_005477.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009981.hg.1 NM_005477.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009981.hg.1 NM_005477.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009981.hg.1 NM_005477.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009981.hg.1 NM_005477.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009981.hg.1 NM_005477.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-79b49c4fc5-ghzkj:80/100.66.129.252:80.
          git-commit: cca8d8ff64ba87988ed84f3fa7b2b2bee312c353
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0