Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009878.hg.1 |
ENST00000412161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009878.hg.1 |
ENST00000412161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009878.hg.1 |
NR_120501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009878.hg.1 |
NR_120501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009878.hg.1 |
NR_120501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009878.hg.1 |
ENST00000412161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009878.hg.1 |
ENST00000412161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009878.hg.1 |
NR_120501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009878.hg.1 |
NR_120501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009878.hg.1 |
NR_120501.1 |
Refseq |