Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04988521_gH
          See other NDUFA4 GE Assays ›
          Gene Symbol:
          NDUFA4
          Gene Name:
          NDUFA4, mitochondrial complex associated
          Gene Aliases:
          CI-9k, CI-MLRQ, MLRQ
          Chromosome Location:
          Chr.7: 10931953 - 10940186 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04988521_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NDUFA4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002489.3 NP_002480.1 2 - 3 263 61
          GenBank mRNA AF201077.1 - 1 187 61
          AK024837.1 - 2 - 3 212 61
          AK130950.1 - 2 - 3 203 61
          BC101794.1 - 2 - 3 187 8069300 61
          BC101796.1 - 2 - 3 187 8069302 61
          BC105295.1 - 2 - 3 234 6739636 61
          BG623521.1 - 264 61
          CR407618.1 - 2 - 3 134 61
          CR541716.1 - 2 - 3 134 61
          U94586.1 - 2 - 3 224 61

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene NDUFA4

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrial electron transport, NADH to ubiquinone
          mitochondrial electron transport, cytochrome c to oxygen
          proton transmembrane transport
          cytochrome-c oxidase activity
          protein binding
          NADH dehydrogenase (ubiquinone) activity
          protein-containing complex binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0900007805.hg.1 ENST00000454946.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010914.hg.1 ENST00000530974.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008662.hg.1 ENST00000505029.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007215.hg.1 ENST00000605507.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007546.hg.1 ENST00000443484.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007546.hg.1 ENST00000443484.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015658.hg.1 ENST00000454121.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014866.hg.1 ENST00000440817.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300008206.hg.1 ENST00000489989.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009309.hg.1 ENST00000416095.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011125.hg.1 ENST00000439946.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010253.hg.1 ENST00000556294.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015479.hg.1 ENST00000448842.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010645.hg.1 ENST00000520009.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011036.hg.1 ENST00000418219.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009882.hg.1 ENST00000450788.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000339600.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000339600.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000339600.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000339600.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000339600.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000339600.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000339600.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000339600.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000339600.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000339600.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000339600.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000339600.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000339600.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000463308.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000463308.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000463308.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000470761.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000470761.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000470761.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000470761.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000470761.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000482299.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000482299.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000482299.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000482299.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000482299.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000482299.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000486007.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000486007.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000486007.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000486007.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000492822.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000492822.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000492822.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000492822.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000492822.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 ENST00000492822.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 NM_002489.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 NM_002489.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 NM_002489.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 NM_002489.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 NM_002489.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 NM_002489.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 NM_002489.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 NM_002489.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 NM_002489.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 NM_002489.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 NM_002489.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010275.hg.1 NM_002489.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010524.hg.1 ENST00000451806.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009398.hg.1 ENST00000485059.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900007805.hg.1 ENST00000454946.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200015658.hg.1 ENST00000454121.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015479.hg.1 ENST00000448842.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300008206.hg.1 ENST00000489989.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010253.hg.1 ENST00000556294.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800010645.hg.1 ENST00000520009.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007215.hg.1 ENST00000605507.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011125.hg.1 ENST00000439946.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007546.hg.1 ENST00000443484.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007546.hg.1 ENST00000443484.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009398.hg.1 ENST00000485059.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010524.hg.1 ENST00000451806.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500008662.hg.1 ENST00000505029.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010914.hg.1 ENST00000530974.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014866.hg.1 ENST00000440817.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100009309.hg.1 ENST00000416095.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000339600.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000339600.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000339600.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000339600.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000339600.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000339600.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000339600.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000339600.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000339600.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000339600.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000339600.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000339600.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000339600.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000463308.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000463308.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000463308.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000470761.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000470761.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000470761.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000470761.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000470761.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000482299.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000482299.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000482299.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000482299.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000482299.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000482299.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000486007.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000486007.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000486007.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000486007.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000492822.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000492822.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000492822.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000492822.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000492822.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 ENST00000492822.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 NM_002489.3 Refseq
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 NM_002489.3 Refseq
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 NM_002489.3 Refseq
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 NM_002489.3 Refseq
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 NM_002489.3 Refseq
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 NM_002489.3 Refseq
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 NM_002489.3 Refseq
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 NM_002489.3 Refseq
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 NM_002489.3 Refseq
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 NM_002489.3 Refseq
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 NM_002489.3 Refseq
          Affymetrix Human Clariom D Assay TC0700010275.hg.1 NM_002489.3 Refseq
          Affymetrix Human Clariom D Assay TC1000009882.hg.1 ENST00000450788.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011036.hg.1 ENST00000418219.1 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0