Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006570.hg.1 |
ENST00000382545.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006570.hg.1 |
ENST00000382545.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006570.hg.1 |
ENST00000382545.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006570.hg.1 |
ENST00000382545.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006570.hg.1 |
ENST00000382545.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006570.hg.1 |
ENST00000382545.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006570.hg.1 |
ENST00000382545.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006570.hg.1 |
ENST00000382545.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006570.hg.1 |
ENST00000382545.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006570.hg.1 |
ENST00000541095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006570.hg.1 |
ENST00000541095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006570.hg.1 |
ENST00000541095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006570.hg.1 |
ENST00000543874.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006570.hg.1 |
ENST00000543874.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006570.hg.1 |
NM_000217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006570.hg.1 |
NM_000217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006570.hg.1 |
NM_000217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006570.hg.1 |
NM_000217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006570.hg.1 |
NM_000217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006570.hg.1 |
NM_000217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006570.hg.1 |
NM_000217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006570.hg.1 |
NM_000217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006570.hg.1 |
NM_000217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006570.hg.1 |
ENST00000382545.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006570.hg.1 |
ENST00000382545.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006570.hg.1 |
ENST00000382545.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006570.hg.1 |
ENST00000382545.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006570.hg.1 |
ENST00000382545.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006570.hg.1 |
ENST00000382545.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006570.hg.1 |
ENST00000382545.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006570.hg.1 |
ENST00000382545.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006570.hg.1 |
ENST00000382545.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006570.hg.1 |
ENST00000541095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006570.hg.1 |
ENST00000541095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006570.hg.1 |
ENST00000541095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006570.hg.1 |
ENST00000543874.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006570.hg.1 |
ENST00000543874.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006570.hg.1 |
NM_000217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006570.hg.1 |
NM_000217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006570.hg.1 |
NM_000217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006570.hg.1 |
NM_000217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006570.hg.1 |
NM_000217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006570.hg.1 |
NM_000217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006570.hg.1 |
NM_000217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006570.hg.1 |
NM_000217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006570.hg.1 |
NM_000217.2 |
Refseq |