Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04994252_m1
          See other EYA1 GE Assays ›
          Gene Symbol:
          EYA1
          Gene Name:
          EYA transcriptional coactivator and phosphatase 1
          Gene Aliases:
          BOP, BOR, BOS1, OFC1
          Chromosome Location:
          Chr.8: 71197433 - 71548130 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04994252_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: EYA1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_017013202.1 XP_016868691.1 1 - 2 108 110
          XM_017013206.1 XP_016868695.1 1 - 2 143 110
          XM_017013209.1 XP_016868698.1 1 - 2 125 110
          GenBank mRNA BC035138.1 - 1 - 2 147 110
          BC048982.1 - 1 - 2 141 110

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the XM_017013202.1, XM_017013206.1, XM_017013209.1 transcripts.

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          establishment of mitotic spindle orientation
          metanephros development
          branching involved in ureteric bud morphogenesis
          outflow tract morphogenesis
          double-strand break repair
          transcription, DNA-templated
          pattern specification process
          mesodermal cell fate specification
          sensory perception of sound
          anatomical structure morphogenesis
          response to ionizing radiation
          striated muscle tissue development
          histone dephosphorylation
          protein sumoylation
          cellular protein localization
          establishment or maintenance of apical/basal cell polarity
          peptidyl-tyrosine dephosphorylation
          aorta morphogenesis
          outer ear morphogenesis
          middle ear morphogenesis
          regulation of neuron differentiation
          positive regulation of DNA repair
          positive regulation of Notch signaling pathway
          positive regulation of transcription by RNA polymerase II
          neuron fate specification
          embryonic skeletal system morphogenesis
          semicircular canal morphogenesis
          positive regulation of epithelial cell proliferation
          pharyngeal system development
          lung epithelial cell differentiation
          otic vesicle morphogenesis
          positive regulation of secondary heart field cardioblast proliferation
          cochlea morphogenesis
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          RNA binding
          protein tyrosine phosphatase activity
          protein binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0800012425.hg.1 ENST00000519927.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012425.hg.1 ENST00000519927.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012425.hg.1 ENST00000519927.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012425.hg.1 ENST00000519927.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012425.hg.1 ENST00000519927.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012425.hg.1 ENST00000523987.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012425.hg.1 ENST00000523987.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012425.hg.1 ENST00000523987.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012425.hg.1 ENST00000523987.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800012425.hg.1 ENST00000519927.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800012425.hg.1 ENST00000519927.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800012425.hg.1 ENST00000519927.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800012425.hg.1 ENST00000519927.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800012425.hg.1 ENST00000519927.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800012425.hg.1 ENST00000523987.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800012425.hg.1 ENST00000523987.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800012425.hg.1 ENST00000523987.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800012425.hg.1 ENST00000523987.4 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0