Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04995079_g1
          See other POU5F1B GE Assays ›
          Gene Symbol:
          POU5F1B
          Gene Name:
          POU class 5 homeobox 1B
          Gene Aliases:
          OCT4-PG1, OCT4PG1, OTF3C, OTF3P1, POU5F1P1, POU5F1P4, POU5FLC20, POU5FLC8
          Chromosome Location:
          Chr.8: 127415612 - 127417210 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04995079_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: POU5F1B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001159542.1 NP_001153014.1 1 104
          GenBank mRNA AF268615.1 - 1 104
          AF268618.1 - 1 104
          DQ486513.1 - 1 104
          FM177969.1 - 1 104
          FM177971.1 - 1 104
          FM177972.1 - 1 104
          FM177974.1 - 1 104
          FM177975.1 - 1 104
          FM177976.1 - 1 104
          FM177977.1 - 1 104
          FM177978.1 - 1 104
          FM177979.1 - 1 104
          FM177980.1 - 1 104
          FM177981.1 - 1 104
          FM177982.1 - 1 104
          GU480835.1 - 1 104
          GU480840.1 - 1 104
          GU480841.1 - 1 104
          GU480842.1 - 1 104
          GU480844.1 - 1 104
          GU480845.1 - 1 104
          GU480849.1 - 1 281 105
          GU480850.1 - 1 281 105
          GU480851.1 - 1 281 105
          GU480864.1 - 1 104
          GU480868.1 - 1 104
          GU480878.1 - 1 104
          GU480880.1 - 1 104
          GU480883.1 - 1 104
          GU480884.1 - 1 104
          GU480886.1 - 1 104
          GU480889.1 - 1 104
          GU480896.1 - 1 104
          GU480899.1 - 1 104
          GU480902.1 - 1 104
          GU480905.1 - 1 104
          GU480910.1 - 1 105
          GU480911.1 - 1 104
          GU480912.1 - 1 104
          GU480913.1 - 1 104
          HQ122710.1 - 1 104
          HQ122711.1 - 1 104
          HQ122712.1 - 1 104
          HQ122713.1 - 1 104
          HQ122714.1 - 1 104
          HQ122715.1 - 1 104
          HQ122716.1 - 1 104
          HQ122717.1 - 1 104
          HQ122718.1 - 1 104
          HQ122719.1 - 1 104
          HQ122720.1 - 1 104
          HQ122721.1 - 1 104
          HQ122749.1 - 1 104
          HQ122750.1 - 1 104
          HQ122751.1 - 1 104
          HQ122752.1 - 1 104
          HQ122781.1 - 1 104
          HQ122782.1 - 1 104
          HQ122783.1 - 1 104
          HQ122784.1 - 1 104
          HQ122785.1 - 1 104
          HQ122786.1 - 1 104
          HQ122787.1 - 1 104
          HQ122788.1 - 1 104
          HQ122789.1 - 1 104
          HQ122790.1 - 1 104
          HQ122816.1 - 1 104
          HQ122817.1 - 1 104
          HQ122818.1 - 1 104
          HQ122819.1 - 1 104
          HQ122820.1 - 1 104
          HQ122821.1 - 1 104
          HQ122822.1 - 1 104
          HQ122823.1 - 1 104
          HQ122824.1 - 1 104
          HQ122825.1 - 1 104
          HQ122826.1 - 1 104
          HQ122841.1 - 1 104
          HQ122842.1 - 1 104
          HQ122844.1 - 1 104
          HQ122845.1 - 1 104
          HQ122846.1 - 1 104
          HQ122847.1 - 1 104
          HQ122848.1 - 1 104
          HQ122849.1 - 1 104
          HQ122850.1 - 1 104
          HQ122851.1 - 1 104
          HQ122852.1 - 1 104
          HQ122853.1 - 1 104
          HQ122854.1 - 1 104
          HQ122855.1 - 1 104
          HQ122856.1 - 1 104
          HQ122857.1 - 1 104
          HQ122863.1 - 1 104
          HQ122864.1 - 1 104
          HQ122865.1 - 1 104
          HQ122866.1 - 1 104
          HQ122867.1 - 1 104
          HQ122868.1 - 1 104
          HQ122869.1 - 1 104
          HQ122870.1 - 1 104
          HQ122871.1 - 1 104
          HQ122874.1 - 1 104
          HQ122875.1 - 1 104
          HQ122876.1 - 1 104
          HQ122877.1 - 1 104
          HQ122878.1 - 1 104
          HQ122879.1 - 1 104
          HQ122880.1 - 1 104
          HQ122881.1 - 1 104
          HQ122882.1 - 1 104
          HQ122883.1 - 1 104
          HQ122884.1 - 1 104
          HQ122885.1 - 1 104
          HQ122886.1 - 1 104
          HQ122887.1 - 1 104
          HQ122891.1 - 1 104
          HQ122892.1 - 1 104
          HQ122894.1 - 1 104
          HQ122895.1 - 1 104
          HQ122896.1 - 1 104
          HQ122897.1 - 1 104
          HQ122898.1 - 1 104
          HQ122899.1 - 1 104
          HQ122900.1 - 1 104
          HQ122901.1 - 1 104
          HQ122902.1 - 1 104

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          DNA-binding transcription factor activity
          sequence-specific DNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0800011331.hg.1 ENST00000521750.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800011331.hg.1 ENST00000521750.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800011331.hg.1 ENST00000521750.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800011331.hg.1 ENST00000521750.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800011331.hg.1 ENST00000521750.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800011331.hg.1 ENST00000521750.3 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix