Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006762.hg.1 |
ENST00000340967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006762.hg.1 |
ENST00000340967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006762.hg.1 |
ENST00000340967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006762.hg.1 |
ENST00000340967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006762.hg.1 |
ENST00000340967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006762.hg.1 |
ENST00000340967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006762.hg.1 |
ENST00000340967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006762.hg.1 |
ENST00000340967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006762.hg.1 |
ENST00000340967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006762.hg.1 |
NM_001010887.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006762.hg.1 |
NM_001010887.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006762.hg.1 |
NM_001010887.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006762.hg.1 |
NM_001010887.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006762.hg.1 |
NM_001010887.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006762.hg.1 |
NM_001010887.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006762.hg.1 |
NM_001010887.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006762.hg.1 |
NM_001010887.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006762.hg.1 |
NM_001010887.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006762.hg.1 |
NM_001010887.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006762.hg.1 |
NM_001010887.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006762.hg.1 |
NM_001010887.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006762.hg.1 |
ENST00000340967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006762.hg.1 |
ENST00000340967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006762.hg.1 |
ENST00000340967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006762.hg.1 |
ENST00000340967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006762.hg.1 |
ENST00000340967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006762.hg.1 |
ENST00000340967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006762.hg.1 |
ENST00000340967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006762.hg.1 |
ENST00000340967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006762.hg.1 |
ENST00000340967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006762.hg.1 |
NM_001010887.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006762.hg.1 |
NM_001010887.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006762.hg.1 |
NM_001010887.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006762.hg.1 |
NM_001010887.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006762.hg.1 |
NM_001010887.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006762.hg.1 |
NM_001010887.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006762.hg.1 |
NM_001010887.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006762.hg.1 |
NM_001010887.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006762.hg.1 |
NM_001010887.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006762.hg.1 |
NM_001010887.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006762.hg.1 |
NM_001010887.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006762.hg.1 |
NM_001010887.2 |
Refseq |