Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04996730_g1
          See other CCL19 GE Assays ›
          Gene Symbol:
          CCL19
          Gene Name:
          C-C motif chemokine ligand 19
          Gene Aliases:
          CKb11, ELC, MIP-3b, MIP3B, SCYA19
          Chromosome Location:
          Chr.9: 34689570 - 34691277 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04996730_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CCL19
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_006274.2 NP_006265.1 2 - 3 320 111
          GenBank mRNA AB000887.1 - 2 - 3 320 111
          AK311945.1 - 2 - 3 319 111
          BC027968.1 - 2 - 3 306 5225849 111
          CR456868.1 - 2 - 3 182 111
          U77180.1 - 2 - 3 182 111
          U88321.1 - 2 - 3 301 111

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CCL19

          Gene Ontology Categories:

          Function(s) Process(es)

          establishment of T cell polarity
          immunological synapse formation
          dendritic cell chemotaxis
          myeloid dendritic cell chemotaxis
          monocyte chemotaxis
          positive regulation of dendritic cell antigen processing and presentation
          cellular calcium ion homeostasis
          inflammatory response
          immune response
          cell communication
          G protein-coupled receptor signaling pathway
          response to virus
          positive regulation of glycoprotein biosynthetic process
          T cell costimulation
          interleukin-12 production
          positive regulation of interleukin-12 production
          positive regulation of tumor necrosis factor production
          response to prostaglandin E
          positive regulation of T cell proliferation
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of GTPase activity
          positive regulation of phosphatidylinositol 3-kinase activity
          positive regulation of T-helper 1 cell differentiation
          positive regulation of endocytosis
          positive regulation of protein kinase activity
          positive regulation of JNK cascade
          lymphocyte chemotaxis
          positive regulation of receptor-mediated endocytosis
          cell maturation
          positive regulation of chemotaxis
          release of sequestered calcium ion into cytosol
          positive regulation of protein kinase B signaling
          regulation of cell projection assembly
          chemokine-mediated signaling pathway
          positive regulation of ERK1 and ERK2 cascade
          cellular response to interferon-gamma
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          response to nitric oxide
          positive regulation of neutrophil chemotaxis
          mature conventional dendritic cell differentiation
          cellular response to virus
          positive regulation of NIK/NF-kappaB signaling
          negative regulation of leukocyte apoptotic process
          positive regulation of cell motility
          positive regulation of dendritic cell dendrite assembly
          chemokine activity
          CCR7 chemokine receptor binding
          CCR10 chemokine receptor binding
          chemokine receptor binding
          CCR chemokine receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0900009915.hg.1 ENST00000311925.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009915.hg.1 ENST00000311925.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009915.hg.1 ENST00000311925.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009915.hg.1 ENST00000311925.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009915.hg.1 ENST00000311925.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009915.hg.1 ENST00000311925.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009915.hg.1 ENST00000311925.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009915.hg.1 ENST00000378800.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009915.hg.1 ENST00000378800.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009915.hg.1 ENST00000378800.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009915.hg.1 ENST00000378800.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009915.hg.1 ENST00000378800.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009915.hg.1 ENST00000378800.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009915.hg.1 ENST00000485502.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009915.hg.1 ENST00000485502.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009915.hg.1 ENST00000485502.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009915.hg.1 ENST00000485502.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009915.hg.1 NM_006274.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009915.hg.1 NM_006274.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009915.hg.1 NM_006274.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009915.hg.1 NM_006274.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009915.hg.1 NM_006274.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009915.hg.1 NM_006274.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009915.hg.1 NM_006274.2 Refseq
          Affymetrix Human Clariom D Assay TC0900009915.hg.1 ENST00000311925.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900009915.hg.1 ENST00000311925.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900009915.hg.1 ENST00000311925.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900009915.hg.1 ENST00000311925.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900009915.hg.1 ENST00000311925.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900009915.hg.1 ENST00000311925.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900009915.hg.1 ENST00000311925.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900009915.hg.1 ENST00000378800.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900009915.hg.1 ENST00000378800.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900009915.hg.1 ENST00000378800.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900009915.hg.1 ENST00000378800.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900009915.hg.1 ENST00000378800.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900009915.hg.1 ENST00000378800.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900009915.hg.1 ENST00000485502.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900009915.hg.1 ENST00000485502.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900009915.hg.1 ENST00000485502.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900009915.hg.1 ENST00000485502.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900009915.hg.1 NM_006274.2 Refseq
          Affymetrix Human Clariom D Assay TC0900009915.hg.1 NM_006274.2 Refseq
          Affymetrix Human Clariom D Assay TC0900009915.hg.1 NM_006274.2 Refseq
          Affymetrix Human Clariom D Assay TC0900009915.hg.1 NM_006274.2 Refseq
          Affymetrix Human Clariom D Assay TC0900009915.hg.1 NM_006274.2 Refseq
          Affymetrix Human Clariom D Assay TC0900009915.hg.1 NM_006274.2 Refseq
          Affymetrix Human Clariom D Assay TC0900009915.hg.1 NM_006274.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0