Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010068.hg.1 |
ENST00000564205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010068.hg.1 |
ENST00000564205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010068.hg.1 |
ENST00000564205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010068.hg.1 |
ENST00000568553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
ENST00000339399.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
ENST00000339399.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
ENST00000339399.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
ENST00000339399.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
ENST00000339399.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
ENST00000339399.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
ENST00000339399.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
ENST00000339399.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
ENST00000339399.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
ENST00000339399.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
ENST00000339399.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
ENST00000339399.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
ENST00000473205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
ENST00000473205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
ENST00000473205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
ENST00000473205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
ENST00000473205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
ENST00000473205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
ENST00000473205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
ENST00000486769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
ENST00000486769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
ENST00000486769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
NM_006401.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
NM_006401.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
NM_006401.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
NM_006401.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
NM_006401.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
NM_006401.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
NM_006401.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
NM_006401.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
NM_006401.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
NM_006401.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
NM_006401.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
NM_006401.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008173.hg.1 |
NM_006401.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
ENST00000339399.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
ENST00000339399.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
ENST00000339399.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
ENST00000339399.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
ENST00000339399.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
ENST00000339399.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
ENST00000339399.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
ENST00000339399.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
ENST00000339399.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
ENST00000339399.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
ENST00000339399.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
ENST00000339399.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
ENST00000473205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
ENST00000473205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
ENST00000473205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
ENST00000473205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
ENST00000473205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
ENST00000473205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
ENST00000473205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
ENST00000486769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
ENST00000486769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
ENST00000486769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
NM_006401.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
NM_006401.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
NM_006401.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
NM_006401.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
NM_006401.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
NM_006401.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
NM_006401.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
NM_006401.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
NM_006401.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
NM_006401.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
NM_006401.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
NM_006401.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008173.hg.1 |
NM_006401.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010068.hg.1 |
ENST00000564205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010068.hg.1 |
ENST00000564205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010068.hg.1 |
ENST00000564205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010068.hg.1 |
ENST00000568553.1 |
Ensembl |